Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 66500261-66500486 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RPL4
Entrez Gene ID
6124
Gene Name
ribosomal protein L4
Gene Aliases
L4
UniGene
Hs.186350
Gene Chromosome Location
Chr.15: 66499315-66504855 on Build GRCh38
Transcripts
NM_000968.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6077490 c.895C>A p.Q299K Chr.15: 66500315-66500315 on Build GRCh38
6263339 c.855T>C p.I285I Chr.15: 66500355-66500355 on Build GRCh38
3503335 c.885C>T p.S295S Chr.15: 66500325-66500325 on Build GRCh38
6769253 c.916C>T p.R306C Chr.15: 66500294-66500294 on Build GRCh38
964237 c.851T>C p.M284T Chr.15: 66500359-66500359 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs897082756 Chr.15: 66500410-66500410 on Build GRCh38
rs751334834 Chr.15: 66500404-66500405 on Build GRCh38
rs754384992 Chr.15: 66500322-66500322 on Build GRCh38
rs978989586 Chr.15: 66500339-66500339 on Build GRCh38
rs759121204 Chr.15: 66500307-66500307 on Build GRCh38
rs755096673 Chr.15: 66500382-66500382 on Build GRCh38
rs777760107 Chr.15: 66500392-66500392 on Build GRCh38
rs200653837 Chr.15: 66500308-66500308 on Build GRCh38
rs750617196 Chr.15: 66500340-66500340 on Build GRCh38
rs977524709 Chr.15: 66500347-66500347 on Build GRCh38
rs780976707 Chr.15: 66500383-66500383 on Build GRCh38
rs149553456 Chr.15: 66500294-66500294 on Build GRCh38
rs765751540 Chr.15: 66500413-66500413 on Build GRCh38
rs758563596 Chr.15: 66500355-66500355 on Build GRCh38
rs765626979 Chr.15: 66500335-66500335 on Build GRCh38
rs749300008 Chr.15: 66500441-66500441 on Build GRCh38
rs895835127 Chr.15: 66500450-66500450 on Build GRCh38
rs749118878 Chr.15: 66500397-66500397 on Build GRCh38
rs1803775 Chr.15: 66500326-66500326 on Build GRCh38
rs762354718 Chr.15: 66500412-66500412 on Build GRCh38
rs748165237 Chr.15: 66500389-66500389 on Build GRCh38
rs764445070 Chr.15: 66500311-66500311 on Build GRCh38
rs78704418 Chr.15: 66500400-66500400 on Build GRCh38
rs1057147227 Chr.15: 66500444-66500444 on Build GRCh38
rs955662616 Chr.15: 66500353-66500353 on Build GRCh38
rs28706938 Chr.15: 66500423-66500423 on Build GRCh38
rs368526038 Chr.15: 66500399-66500399 on Build GRCh38
rs763311785 Chr.15: 66500424-66500424 on Build GRCh38
rs780353785 Chr.15: 66500369-66500369 on Build GRCh38
rs111585548 Chr.15: 66500292-66500292 on Build GRCh38
rs751525940 Chr.15: 66500380-66500380 on Build GRCh38
rs769682672 Chr.15: 66500391-66500391 on Build GRCh38
rs941234357 Chr.15: 66500403-66500403 on Build GRCh38
rs375190154 Chr.15: 66500407-66500407 on Build GRCh38
rs371607586 Chr.15: 66500401-66500401 on Build GRCh38

View Full Product Details