Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 28793676-28793915 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PPP1CB
Entrez Gene ID
5500
Gene Name
protein phosphatase 1 catalytic subunit beta
Gene Aliases
HEL-S-80p, PP-1B, PP1B, PP1beta, PPP1CD
UniGene
Hs.702907
Gene Chromosome Location
Chr.2: 28751748-28802940 on Build GRCh38
Transcripts
NM_002709.2 NM_206876.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6247813 c.747G>T p.V249V Chr.2: 28793865-28793865 on Build GRCh38
6753785 c.793T>G p.L265V Chr.2: 28793911-28793911 on Build GRCh38
6467171 c.778C>T p.R260* Chr.2: 28793896-28793896 on Build GRCh38
23148 c.748G>A p.V250M Chr.2: 28793866-28793866 on Build GRCh38
345314 c.752A>G p.E251G Chr.2: 28793870-28793870 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs765535212 Chr.2: 28793825-28793825 on Build GRCh38
rs886037953 Chr.2: 28793872-28793872 on Build GRCh38
rs1031295046 Chr.2: 28793777-28793777 on Build GRCh38
rs993064254 Chr.2: 28793811-28793811 on Build GRCh38
rs776487891 Chr.2: 28793712-28793712 on Build GRCh38
rs777536645 Chr.2: 28793889-28793889 on Build GRCh38
rs943165171 Chr.2: 28793768-28793768 on Build GRCh38
rs758258613 Chr.2: 28793832-28793832 on Build GRCh38
rs754933712 Chr.2: 28793853-28793853 on Build GRCh38
rs578033831 Chr.2: 28793725-28793725 on Build GRCh38
rs747736240 Chr.2: 28793858-28793858 on Build GRCh38
rs755702703 Chr.2: 28793860-28793860 on Build GRCh38
rs556430840 Chr.2: 28793711-28793711 on Build GRCh38
rs955904253 Chr.2: 28793789-28793789 on Build GRCh38
rs780531331 Chr.2: 28793854-28793854 on Build GRCh38
rs1040491757 Chr.2: 28793774-28793774 on Build GRCh38
rs201590987 Chr.2: 28793821-28793821 on Build GRCh38
rs750206948 Chr.2: 28793828-28793828 on Build GRCh38
rs766166974 Chr.2: 28793845-28793845 on Build GRCh38
rs547821228 Chr.2: 28793772-28793773 on Build GRCh38
rs1017420488 Chr.2: 28793736-28793736 on Build GRCh38
rs751441010 Chr.2: 28793848-28793848 on Build GRCh38
rs910381283 Chr.2: 28793732-28793732 on Build GRCh38

View Full Product Details