Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 57646980-57647217 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
YPEL4
Entrez Gene ID
219539
Gene Name
yippee like 4
Gene Aliases
-
UniGene
Hs.135978
Gene Chromosome Location
Chr.11: 57645087-57649944 on Build GRCh38
Transcripts
NM_145008.2 XM_017017318.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6200969 c.66G>A p.L22L Chr.11: 57647042-57647042 on Build GRCh38
5812786 c.91T>A p.C31S Chr.11: 57647017-57647017 on Build GRCh38
3788004 c.29C>A p.P10H Chr.11: 57647079-57647079 on Build GRCh38
1354797 c.42_43insC p.T15fs*29 Chr.11: 57647065-57647066 on Build GRCh38
233783 c.71G>A p.R24H Chr.11: 57647037-57647037 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1057013179 Chr.11: 57647089-57647089 on Build GRCh38
rs1007023322 Chr.11: 57647172-57647172 on Build GRCh38
rs768968117 Chr.11: 57647127-57647127 on Build GRCh38
rs762423127 Chr.11: 57647171-57647171 on Build GRCh38
rs958105426 Chr.11: 57647191-57647191 on Build GRCh38
rs1025124277 Chr.11: 57647107-57647107 on Build GRCh38
rs904260175 Chr.11: 57647120-57647120 on Build GRCh38
rs780232043 Chr.11: 57647065-57647065 on Build GRCh38
rs937168776 Chr.11: 57647087-57647087 on Build GRCh38
rs774711134 Chr.11: 57647183-57647183 on Build GRCh38
rs759180777 Chr.11: 57647121-57647121 on Build GRCh38
rs746309170 Chr.11: 57647052-57647052 on Build GRCh38
rs779597989 Chr.11: 57647037-57647038 on Build GRCh38
rs555492094 Chr.11: 57647173-57647173 on Build GRCh38
rs368689970 Chr.11: 57647153-57647153 on Build GRCh38
rs764311509 Chr.11: 57647147-57647147 on Build GRCh38
rs945727650 Chr.11: 57647096-57647096 on Build GRCh38
rs757763202 Chr.11: 57647041-57647041 on Build GRCh38
rs1032824714 Chr.11: 57647170-57647170 on Build GRCh38
rs375204995 Chr.11: 57647085-57647085 on Build GRCh38
rs73480570 Chr.11: 57647110-57647110 on Build GRCh38
rs201676800 Chr.11: 57647150-57647150 on Build GRCh38
rs773167878 Chr.11: 57647106-57647106 on Build GRCh38
rs751196775 Chr.11: 57647004-57647004 on Build GRCh38
rs758581663 Chr.11: 57647053-57647053 on Build GRCh38
rs372464629 Chr.11: 57647111-57647111 on Build GRCh38
rs767186910 Chr.11: 57647145-57647145 on Build GRCh38
rs754227451 Chr.11: 57647037-57647037 on Build GRCh38
rs773881385 Chr.11: 57647114-57647114 on Build GRCh38
rs868701033 Chr.11: 57647028-57647028 on Build GRCh38
rs370790912 Chr.11: 57647011-57647011 on Build GRCh38
rs769514955 Chr.11: 57647090-57647090 on Build GRCh38
rs768887913 Chr.11: 57647084-57647084 on Build GRCh38
rs376976563 Chr.11: 57647005-57647005 on Build GRCh38
rs779575005 Chr.11: 57647046-57647046 on Build GRCh38
rs572409258 Chr.11: 57647034-57647034 on Build GRCh38
rs747240932 Chr.11: 57647083-57647083 on Build GRCh38

View Full Product Details