Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 124451554-124451823 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TRMT12
Entrez Gene ID
55039
Gene Name
tRNA methyltransferase 12 homolog
Gene Aliases
TRM12, TYW2
UniGene
Hs.9925
Gene Chromosome Location
Chr.8: 124450807-124453026 on Build GRCh38
Transcripts
NM_017956.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5782276 c.721C>A p.L241I Chr.8: 124451648-124451648 on Build GRCh38
5789066 c.638C>T p.T213I Chr.8: 124451565-124451565 on Build GRCh38
6499993 c.853G>C p.D285H Chr.8: 124451780-124451780 on Build GRCh38
277818 c.884G>T p.R295I Chr.8: 124451811-124451811 on Build GRCh38
5062871 c.699T>C p.C233C Chr.8: 124451626-124451626 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs778004728 Chr.8: 124451602-124451602 on Build GRCh38
rs769625192 Chr.8: 124451607-124451607 on Build GRCh38
rs530429825 Chr.8: 124451704-124451704 on Build GRCh38
rs758731270 Chr.8: 124451591-124451591 on Build GRCh38
rs767344232 Chr.8: 124451655-124451655 on Build GRCh38
rs544340342 Chr.8: 124451623-124451623 on Build GRCh38
rs759690922 Chr.8: 124451584-124451585 on Build GRCh38
rs745525280 Chr.8: 124451603-124451603 on Build GRCh38
rs775114934 Chr.8: 124451616-124451616 on Build GRCh38
rs867273537 Chr.8: 124451712-124451712 on Build GRCh38
rs1050325334 Chr.8: 124451783-124451783 on Build GRCh38
rs748964795 Chr.8: 124451780-124451780 on Build GRCh38
rs1017909875 Chr.8: 124451609-124451609 on Build GRCh38
rs928518079 Chr.8: 124451718-124451718 on Build GRCh38
rs777835643 Chr.8: 124451784-124451784 on Build GRCh38
rs772938177 Chr.8: 124451679-124451679 on Build GRCh38
rs148858836 Chr.8: 124451696-124451696 on Build GRCh38
rs928004887 Chr.8: 124451660-124451660 on Build GRCh38
rs761664348 Chr.8: 124451647-124451647 on Build GRCh38
rs753180790 Chr.8: 124451731-124451731 on Build GRCh38
rs755794960 Chr.8: 124451765-124451765 on Build GRCh38
rs754092053 Chr.8: 124451697-124451697 on Build GRCh38
rs200399722 Chr.8: 124451788-124451788 on Build GRCh38
rs946622341 Chr.8: 124451751-124451751 on Build GRCh38
rs377165981 Chr.8: 124451787-124451787 on Build GRCh38
rs142387012 Chr.8: 124451743-124451743 on Build GRCh38
rs768306757 Chr.8: 124451622-124451622 on Build GRCh38
rs373767717 Chr.8: 124451733-124451733 on Build GRCh38
rs748624577 Chr.8: 124451589-124451589 on Build GRCh38
rs760866071 Chr.8: 124451680-124451680 on Build GRCh38
rs765462553 Chr.8: 124451656-124451656 on Build GRCh38
rs778181703 Chr.8: 124451738-124451738 on Build GRCh38
rs138956356 Chr.8: 124451729-124451729 on Build GRCh38
rs199844898 Chr.8: 124451617-124451617 on Build GRCh38
rs372554468 Chr.8: 124451769-124451769 on Build GRCh38
rs753060404 Chr.8: 124451696-124451697 on Build GRCh38
rs981320636 Chr.8: 124451708-124451708 on Build GRCh38

View Full Product Details