Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.21: 45265275-45265528 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
POFUT2
Entrez Gene ID
23275
Gene Name
protein O-fucosyltransferase 2
Gene Aliases
C21orf80, FUT13
UniGene
Hs.592164
Gene Chromosome Location
Chr.21: 45263928-45287912 on Build GRCh38
Transcripts
NM_015227.4 NM_133635.4 NR_004858.1 XM_006723981.3 XM_011529504.2 XM_011529506.2 XM_011529507.2 XM_011529510.2 XM_017028306.1 XM_017028309.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6267551 c.1266C>A p.P422P Chr.21: 45265506-45265506 on Build GRCh38
4943640 c.1285T>G p.Y429D Chr.21: 45265487-45265487 on Build GRCh38
6004690 c.1253C>T p.A418V Chr.21: 45265519-45265519 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs374968449 Chr.21: 45265437-45265437 on Build GRCh38
rs565037703 Chr.21: 45265464-45265464 on Build GRCh38
rs778469737 Chr.21: 45265439-45265439 on Build GRCh38
rs368685176 Chr.21: 45265451-45265451 on Build GRCh38
rs58387020 Chr.21: 45265441-45265441 on Build GRCh38
rs549487953 Chr.21: 45265316-45265316 on Build GRCh38
rs767430817 Chr.21: 45265506-45265506 on Build GRCh38
rs956661387 Chr.21: 45265504-45265504 on Build GRCh38
rs199690917 Chr.21: 45265456-45265456 on Build GRCh38
rs1001302516 Chr.21: 45265488-45265488 on Build GRCh38
rs540518453 Chr.21: 45265457-45265457 on Build GRCh38
rs527441350 Chr.21: 45265386-45265386 on Build GRCh38
rs980932770 Chr.21: 45265388-45265388 on Build GRCh38
rs908147244 Chr.21: 45265418-45265418 on Build GRCh38
rs944976424 Chr.21: 45265419-45265419 on Build GRCh38
rs553428556 Chr.21: 45265357-45265357 on Build GRCh38
rs756770316 Chr.21: 45265460-45265460 on Build GRCh38
rs533319429 Chr.21: 45265477-45265477 on Build GRCh38
rs919578024 Chr.21: 45265394-45265394 on Build GRCh38
rs182942870 Chr.21: 45265317-45265317 on Build GRCh38
rs910521928 Chr.21: 45265416-45265416 on Build GRCh38
rs556606393 Chr.21: 45265383-45265383 on Build GRCh38
rs577561643 Chr.21: 45265366-45265366 on Build GRCh38
rs768056133 Chr.21: 45265446-45265446 on Build GRCh38
rs758049666 Chr.21: 45265452-45265452 on Build GRCh38
rs374080840 Chr.21: 45265408-45265408 on Build GRCh38
rs762108988 Chr.21: 45265503-45265503 on Build GRCh38
rs567886168 Chr.21: 45265335-45265335 on Build GRCh38
rs961162399 Chr.21: 45265382-45265382 on Build GRCh38
rs930702952 Chr.21: 45265407-45265407 on Build GRCh38
rs570171977 Chr.21: 45265294-45265294 on Build GRCh38
rs367592691 Chr.21: 45265448-45265448 on Build GRCh38
rs991977493 Chr.21: 45265376-45265376 on Build GRCh38
rs115550681 Chr.21: 45265509-45265509 on Build GRCh38
rs75274352 Chr.21: 45265336-45265336 on Build GRCh38
rs780764583 Chr.21: 45265462-45265462 on Build GRCh38
rs147423047 Chr.21: 45265494-45265494 on Build GRCh38
rs115440615 Chr.21: 45265381-45265381 on Build GRCh38
rs768890239 Chr.21: 45265489-45265489 on Build GRCh38
rs1012994828 Chr.21: 45265331-45265331 on Build GRCh38
rs374936952 Chr.21: 45265444-45265444 on Build GRCh38
rs751636139 Chr.21: 45265443-45265443 on Build GRCh38
rs769652085 Chr.21: 45265433-45265433 on Build GRCh38
rs372678621 Chr.21: 45265422-45265422 on Build GRCh38
rs114378659 Chr.21: 45265310-45265310 on Build GRCh38
rs745730019 Chr.21: 45265463-45265463 on Build GRCh38
rs753506266 Chr.21: 45265297-45265297 on Build GRCh38
rs372886985 Chr.21: 45265450-45265450 on Build GRCh38
rs370486874 Chr.21: 45265442-45265442 on Build GRCh38
rs775374899 Chr.21: 45265468-45265468 on Build GRCh38
rs930986172 Chr.21: 45265399-45265399 on Build GRCh38
rs756925203 Chr.21: 45265365-45265365 on Build GRCh38
rs901965010 Chr.21: 45265298-45265298 on Build GRCh38

View Full Product Details