Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 137234315-137234542 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SLC34A3
Entrez Gene ID
142680
Gene Name
solute carrier family 34 member 3
Gene Aliases
HHRH, NPTIIc
UniGene
Hs.432442
Gene Chromosome Location
Chr.9: 137230757-137236554 on Build GRCh38
Transcripts
NM_001177316.1 NM_001177317.1 NM_080877.2 XM_011518257.2 XM_011518258.1 XM_011518259.1 XM_011518260.1 XM_011518261.2 XM_017014290.1 XM_017014291.1 XM_017014292.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6781568 c.1201C>T p.P401S Chr.9: 137234523-137234523 on Build GRCh38
6048060 c.1167G>A p.L389L Chr.9: 137234489-137234489 on Build GRCh38
2151979 c.1187C>T p.T396M Chr.9: 137234509-137234509 on Build GRCh38
3716263 c.1192G>A p.A398T Chr.9: 137234514-137234514 on Build GRCh38
4850478 c.1140C>T p.L380L Chr.9: 137234462-137234462 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs370453108 Chr.9: 137234393-137234393 on Build GRCh38
rs775994027 Chr.9: 137234427-137234427 on Build GRCh38
rs752540173 Chr.9: 137234383-137234383 on Build GRCh38
rs1004262765 Chr.9: 137234493-137234493 on Build GRCh38
rs970151443 Chr.9: 137234420-137234420 on Build GRCh38
rs1000009986 Chr.9: 137234344-137234344 on Build GRCh38
rs368525218 Chr.9: 137234457-137234457 on Build GRCh38
rs748862410 Chr.9: 137234520-137234520 on Build GRCh38
rs995324067 Chr.9: 137234401-137234401 on Build GRCh38
rs761490521 Chr.9: 137234524-137234524 on Build GRCh38
rs545423705 Chr.9: 137234350-137234350 on Build GRCh38
rs772448315 Chr.9: 137234412-137234412 on Build GRCh38
rs374371470 Chr.9: 137234402-137234402 on Build GRCh38
rs201720181 Chr.9: 137234519-137234519 on Build GRCh38
rs770087210 Chr.9: 137234403-137234403 on Build GRCh38
rs748479353 Chr.9: 137234411-137234411 on Build GRCh38
rs141417134 Chr.9: 137234510-137234510 on Build GRCh38
rs765546536 Chr.9: 137234441-137234443 on Build GRCh38
rs779999649 Chr.9: 137234397-137234397 on Build GRCh38
rs766680009 Chr.9: 137234456-137234456 on Build GRCh38
rs377197436 Chr.9: 137234390-137234390 on Build GRCh38
rs768436440 Chr.9: 137234487-137234487 on Build GRCh38
rs774001691 Chr.9: 137234492-137234492 on Build GRCh38
rs35535797 Chr.9: 137234406-137234406 on Build GRCh38
rs755322848 Chr.9: 137234419-137234420 on Build GRCh38
rs766479116 Chr.9: 137234502-137234502 on Build GRCh38
rs113568956 Chr.9: 137234462-137234462 on Build GRCh38
rs369565150 Chr.9: 137234465-137234465 on Build GRCh38
rs757694011 Chr.9: 137234515-137234515 on Build GRCh38
rs201401168 Chr.9: 137234472-137234472 on Build GRCh38
rs199701166 Chr.9: 137234513-137234513 on Build GRCh38
rs770417865 Chr.9: 137234422-137234422 on Build GRCh38
rs745352438 Chr.9: 137234482-137234482 on Build GRCh38
rs201759654 Chr.9: 137234445-137234445 on Build GRCh38
rs762986714 Chr.9: 137234486-137234486 on Build GRCh38
rs764008678 Chr.9: 137234444-137234444 on Build GRCh38
rs759408375 Chr.9: 137234366-137234366 on Build GRCh38
rs560440785 Chr.9: 137234505-137234505 on Build GRCh38
rs199536442 Chr.9: 137234471-137234471 on Build GRCh38
rs565479962 Chr.9: 137234468-137234468 on Build GRCh38
rs374826755 Chr.9: 137234464-137234464 on Build GRCh38
rs759155749 Chr.9: 137234428-137234428 on Build GRCh38
rs775620578 Chr.9: 137234441-137234441 on Build GRCh38
rs550877277 Chr.9: 137234429-137234429 on Build GRCh38
rs150835225 Chr.9: 137234512-137234512 on Build GRCh38
rs1027882028 Chr.9: 137234423-137234423 on Build GRCh38
rs762784419 Chr.9: 137234442-137234442 on Build GRCh38
rs894530845 Chr.9: 137234356-137234356 on Build GRCh38
rs745543829 Chr.9: 137234517-137234517 on Build GRCh38
rs922090983 Chr.9: 137234473-137234473 on Build GRCh38
rs200677629 Chr.9: 137234501-137234501 on Build GRCh38
rs779344191 Chr.9: 137234463-137234463 on Build GRCh38
rs1022794461 Chr.9: 137234516-137234516 on Build GRCh38
rs756300584 Chr.9: 137234454-137234454 on Build GRCh38
rs769447120 Chr.9: 137234483-137234483 on Build GRCh38
rs922261737 Chr.9: 137234452-137234452 on Build GRCh38
rs890746190 Chr.9: 137234357-137234357 on Build GRCh38
rs765325483 Chr.9: 137234367-137234367 on Build GRCh38
rs201683019 Chr.9: 137234484-137234484 on Build GRCh38
rs758312323 Chr.9: 137234386-137234386 on Build GRCh38
rs768473327 Chr.9: 137234523-137234523 on Build GRCh38
rs746648106 Chr.9: 137234418-137234418 on Build GRCh38
rs138798032 Chr.9: 137234509-137234509 on Build GRCh38
rs200481736 Chr.9: 137234417-137234417 on Build GRCh38
rs1015897530 Chr.9: 137234372-137234372 on Build GRCh38

View Full Product Details