Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 38374907-38375180 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
XYLB
Entrez Gene ID
9942
Gene Name
xylulokinase
Gene Aliases
-
UniGene
Hs.659985
Gene Chromosome Location
Chr.3: 38346760-38453041 on Build GRCh38
Transcripts
NM_005108.3 XM_011534325.2 XM_011534327.1 XM_011534328.2 XM_011534329.1 XM_011534330.2 XM_017007595.1 XM_017007596.1 XM_017007597.1 XM_017007598.1 XM_017007599.1 XM_017007600.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3392373 c.891C>T p.V297V Chr.3: 38375146-38375146 on Build GRCh38
6428578 c.914T>C p.L305P Chr.3: 38375169-38375169 on Build GRCh38
6479767 c.895C>T p.L299L Chr.3: 38375150-38375150 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs758273105 Chr.3: 38375066-38375066 on Build GRCh38
rs766034404 Chr.3: 38375102-38375102 on Build GRCh38
rs778191960 Chr.3: 38375130-38375130 on Build GRCh38
rs372458925 Chr.3: 38375020-38375020 on Build GRCh38
rs1001570655 Chr.3: 38375106-38375106 on Build GRCh38
rs112046512 Chr.3: 38375076-38375076 on Build GRCh38
rs921731209 Chr.3: 38374974-38374974 on Build GRCh38
rs756435565 Chr.3: 38375127-38375127 on Build GRCh38
rs905896230 Chr.3: 38375105-38375105 on Build GRCh38
rs368035732 Chr.3: 38375096-38375096 on Build GRCh38
rs753186765 Chr.3: 38375126-38375126 on Build GRCh38
rs959492036 Chr.3: 38375087-38375087 on Build GRCh38
rs1045833036 Chr.3: 38375075-38375075 on Build GRCh38
rs374400841 Chr.3: 38374988-38374988 on Build GRCh38
rs995003506 Chr.3: 38375120-38375120 on Build GRCh38
rs1003721052 Chr.3: 38375077-38375077 on Build GRCh38
rs754789255 Chr.3: 38375038-38375038 on Build GRCh38
rs948031647 Chr.3: 38375074-38375074 on Build GRCh38
rs1010797509 Chr.3: 38375141-38375141 on Build GRCh38
rs749567221 Chr.3: 38375132-38375132 on Build GRCh38
rs1033543603 Chr.3: 38375108-38375108 on Build GRCh38
rs935824460 Chr.3: 38375030-38375030 on Build GRCh38
rs576551695 Chr.3: 38375079-38375079 on Build GRCh38
rs754496383 Chr.3: 38375115-38375115 on Build GRCh38
rs542197559 Chr.3: 38375081-38375081 on Build GRCh38
rs374939738 Chr.3: 38375095-38375095 on Build GRCh38
rs1033900626 Chr.3: 38375080-38375080 on Build GRCh38
rs751080838 Chr.3: 38375104-38375104 on Build GRCh38
rs745982811 Chr.3: 38375151-38375151 on Build GRCh38
rs983451506 Chr.3: 38375134-38375134 on Build GRCh38
rs546114134 Chr.3: 38375052-38375052 on Build GRCh38
rs779008197 Chr.3: 38375146-38375146 on Build GRCh38
rs781572444 Chr.3: 38375123-38375123 on Build GRCh38

View Full Product Details