Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 44374029-44374276 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SERP2
Entrez Gene ID
387923
Gene Name
stress-associated endoplasmic reticulum protein family member 2
Gene Aliases
C13orf21, bA269C23.1
UniGene
Hs.377972
Gene Chromosome Location
Chr.13: 44373842-44397714 on Build GRCh38
Transcripts
NM_001010897.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1366992 c.33C>T p.N11N Chr.13: 44374058-44374058 on Build GRCh38
4845880 c.60G>A p.Q20Q Chr.13: 44374085-44374085 on Build GRCh38
5002151 c.69C>A p.N23K Chr.13: 44374094-44374094 on Build GRCh38
6463229 c.61A>G p.R21G Chr.13: 44374086-44374086 on Build GRCh38
1265275 c.80C>T p.T27I Chr.13: 44374105-44374105 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs779769478 Chr.13: 44374142-44374142 on Build GRCh38
rs765366799 Chr.13: 44374093-44374093 on Build GRCh38
rs531107294 Chr.13: 44374058-44374058 on Build GRCh38
rs754054255 Chr.13: 44374091-44374091 on Build GRCh38
rs751886993 Chr.13: 44374157-44374168 on Build GRCh38
rs61947535 Chr.13: 44374167-44374167 on Build GRCh38
rs554659975 Chr.13: 44374257-44374257 on Build GRCh38
rs751981720 Chr.13: 44374129-44374129 on Build GRCh38
rs138740370 Chr.13: 44374046-44374046 on Build GRCh38
rs375066449 Chr.13: 44374133-44374133 on Build GRCh38
rs997446876 Chr.13: 44374159-44374159 on Build GRCh38
rs965446763 Chr.13: 44374206-44374206 on Build GRCh38
rs768721176 Chr.13: 44374155-44374155 on Build GRCh38
rs749181723 Chr.13: 44374154-44374154 on Build GRCh38
rs774211589 Chr.13: 44374066-44374066 on Build GRCh38
rs895143837 Chr.13: 44374176-44374176 on Build GRCh38
rs369961302 Chr.13: 44374130-44374130 on Build GRCh38
rs1038385284 Chr.13: 44374221-44374221 on Build GRCh38
rs768558019 Chr.13: 44374062-44374062 on Build GRCh38
rs67144714 Chr.13: 44374164-44374164 on Build GRCh38
rs756735354 Chr.13: 44374092-44374092 on Build GRCh38
rs778043392 Chr.13: 44374116-44374116 on Build GRCh38
rs952830069 Chr.13: 44374253-44374253 on Build GRCh38
rs201663302 Chr.13: 44374159-44374160 on Build GRCh38
rs760743999 Chr.13: 44374082-44374082 on Build GRCh38
rs924963427 Chr.13: 44374171-44374171 on Build GRCh38
rs956663492 Chr.13: 44374177-44374177 on Build GRCh38
rs571158773 Chr.13: 44374109-44374109 on Build GRCh38
rs371733734 Chr.13: 44374131-44374131 on Build GRCh38
rs766645178 Chr.13: 44374087-44374087 on Build GRCh38
rs61947533 Chr.13: 44374164-44374164 on Build GRCh38
rs199705926 Chr.13: 44374167-44374167 on Build GRCh38
rs897203637 Chr.13: 44374238-44374238 on Build GRCh38
rs200371760 Chr.13: 44374094-44374094 on Build GRCh38
rs140520721 Chr.13: 44374060-44374060 on Build GRCh38
rs61947534 Chr.13: 44374165-44374165 on Build GRCh38
rs994790391 Chr.13: 44374214-44374214 on Build GRCh38
rs1025905361 Chr.13: 44374239-44374239 on Build GRCh38

View Full Product Details