Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 121355982-121356229 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
STOM
Entrez Gene ID
2040
Gene Name
stomatin
Gene Aliases
BND7, EPB7, EPB72
UniGene
Hs.253903
Gene Chromosome Location
Chr.9: 121338988-121370304 on Build GRCh38
Transcripts
NM_001270526.1 NM_001270527.1 NM_004099.5 NM_198194.2 NR_073037.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5711130 c.142A>G p.I48V Chr.9: 121356076-121356076 on Build GRCh38
1104776 c.127G>A p.V43I Chr.9: 121356091-121356091 on Build GRCh38
5021628 c.76G>C p.G26R Chr.9: 121356142-121356142 on Build GRCh38
6792178 c.126C>T p.T42T Chr.9: 121356092-121356092 on Build GRCh38
5021629 c.71G>A p.S24N Chr.9: 121356147-121356147 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs779929351 Chr.9: 121356131-121356131 on Build GRCh38
rs774962680 Chr.9: 121356087-121356087 on Build GRCh38
rs1037218769 Chr.9: 121356111-121356111 on Build GRCh38
rs887532236 Chr.9: 121356107-121356107 on Build GRCh38
rs747580083 Chr.9: 121356143-121356143 on Build GRCh38
rs757131877 Chr.9: 121356037-121356037 on Build GRCh38
rs1024727773 Chr.9: 121356186-121356186 on Build GRCh38
rs772570024 Chr.9: 121356173-121356173 on Build GRCh38
rs984833894 Chr.9: 121356154-121356154 on Build GRCh38
rs746816228 Chr.9: 121356133-121356133 on Build GRCh38
rs759993289 Chr.9: 121356164-121356164 on Build GRCh38
rs758512810 Chr.9: 121356045-121356045 on Build GRCh38
rs527633384 Chr.9: 121356009-121356009 on Build GRCh38
rs753860952 Chr.9: 121356112-121356112 on Build GRCh38
rs114133174 Chr.9: 121356110-121356110 on Build GRCh38
rs921640515 Chr.9: 121356035-121356035 on Build GRCh38
rs141204528 Chr.9: 121356140-121356140 on Build GRCh38
rs568321675 Chr.9: 121356091-121356091 on Build GRCh38
rs778697894 Chr.9: 121356124-121356124 on Build GRCh38
rs372101329 Chr.9: 121356106-121356106 on Build GRCh38
rs940393324 Chr.9: 121356195-121356195 on Build GRCh38
rs768429679 Chr.9: 121356070-121356070 on Build GRCh38
rs913127452 Chr.9: 121356002-121356002 on Build GRCh38
rs772045398 Chr.9: 121356054-121356054 on Build GRCh38
rs752962469 Chr.9: 121356021-121356021 on Build GRCh38
rs907551270 Chr.9: 121356183-121356183 on Build GRCh38
rs761097217 Chr.9: 121356177-121356177 on Build GRCh38
rs754141736 Chr.9: 121356192-121356192 on Build GRCh38
rs745622820 Chr.9: 121356048-121356048 on Build GRCh38
rs766965853 Chr.9: 121356121-121356122 on Build GRCh38
rs772615715 Chr.9: 121356156-121356156 on Build GRCh38
rs368376420 Chr.9: 121356074-121356074 on Build GRCh38
rs756347795 Chr.9: 121356023-121356023 on Build GRCh38
rs761967190 Chr.9: 121356193-121356193 on Build GRCh38
rs745770275 Chr.9: 121356127-121356127 on Build GRCh38
rs764194091 Chr.9: 121356182-121356182 on Build GRCh38
rs773775624 Chr.9: 121356147-121356147 on Build GRCh38
rs374202822 Chr.9: 121356026-121356026 on Build GRCh38
rs746620260 Chr.9: 121356063-121356063 on Build GRCh38
rs749387798 Chr.9: 121356034-121356034 on Build GRCh38
rs761443183 Chr.9: 121356076-121356076 on Build GRCh38
rs200987792 Chr.9: 121356137-121356137 on Build GRCh38
rs371879520 Chr.9: 121356015-121356015 on Build GRCh38
rs757402987 Chr.9: 121356115-121356115 on Build GRCh38
rs775771948 Chr.9: 121356176-121356176 on Build GRCh38
rs769394949 Chr.9: 121356148-121356148 on Build GRCh38
rs766789820 Chr.9: 121356077-121356077 on Build GRCh38
rs373417057 Chr.9: 121356062-121356062 on Build GRCh38
rs766503763 Chr.9: 121356092-121356092 on Build GRCh38
rs944639551 Chr.9: 121356024-121356024 on Build GRCh38
rs765320665 Chr.9: 121356198-121356198 on Build GRCh38
rs368984951 Chr.9: 121356128-121356128 on Build GRCh38

View Full Product Details