Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 8419613-8419850 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SLC35B3
Entrez Gene ID
51000
Gene Name
solute carrier family 35 member B3
Gene Aliases
C6orf196, CGI-19, PAPST2
UniGene
Hs.285847
Gene Chromosome Location
Chr.6: 8413066-8435606 on Build GRCh38
Transcripts
NM_001142541.2 NM_015948.4 NR_109913.1 NR_109914.1 XM_005249156.3 XM_005249157.2 XM_005249158.1 XM_006715102.3 XM_006715103.3 XM_011514648.1 XM_011514650.2 XM_017010910.1 XM_017010911.1 XM_017010912.1 XM_017010913.1 XM_017010914.1 XM_017010915.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6447396 c.686T>A p.V229E Chr.6: 8419674-8419674 on Build GRCh38
3176653 c.699C>T p.S233S Chr.6: 8419661-8419661 on Build GRCh38
5006703 c.683-1G>A p.? Chr.6: 8419678-8419678 on Build GRCh38
6447395 c.686T>A p.V229E Chr.6: 8419674-8419674 on Build GRCh38
6396107 c.721G>C p.V241L Chr.6: 8419639-8419639 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs765264359 Chr.6: 8419728-8419728 on Build GRCh38
rs753309521 Chr.6: 8419662-8419662 on Build GRCh38
rs756576208 Chr.6: 8419696-8419696 on Build GRCh38
rs116734551 Chr.6: 8419700-8419700 on Build GRCh38
rs765968010 Chr.6: 8419651-8419651 on Build GRCh38
rs35150610 Chr.6: 8419676-8419676 on Build GRCh38
rs746056642 Chr.6: 8419722-8419722 on Build GRCh38
rs747428042 Chr.6: 8419705-8419705 on Build GRCh38
rs777848986 Chr.6: 8419700-8419700 on Build GRCh38
rs1005553167 Chr.6: 8419805-8419805 on Build GRCh38
rs749629948 Chr.6: 8419708-8419708 on Build GRCh38
rs748132000 Chr.6: 8419692-8419693 on Build GRCh38
rs758025919 Chr.6: 8419647-8419647 on Build GRCh38
rs757569906 Chr.6: 8419711-8419711 on Build GRCh38
rs750043929 Chr.6: 8419639-8419639 on Build GRCh38
rs34032207 Chr.6: 8419659-8419660 on Build GRCh38
rs758664032 Chr.6: 8419697-8419700 on Build GRCh38
rs901366430 Chr.6: 8419675-8419675 on Build GRCh38
rs747031781 Chr.6: 8419729-8419729 on Build GRCh38
rs1034330673 Chr.6: 8419813-8419813 on Build GRCh38
rs1016770733 Chr.6: 8419715-8419715 on Build GRCh38
rs888536846 Chr.6: 8419804-8419804 on Build GRCh38

View Full Product Details