Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 103777896-103778123 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NT5DC3
Entrez Gene ID
51559
Gene Name
5'-nucleotidase domain containing 3
Gene Aliases
TU12B1-TY, TU12B1TY
UniGene
Hs.48428
Gene Chromosome Location
Chr.12: 103746315-103841238 on Build GRCh38
Transcripts
NM_001031701.2 XM_011538476.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4574954 c.1533C>T p.D511D Chr.12: 103777943-103777943 on Build GRCh38
3752852 c.1512G>A p.L504L Chr.12: 103777964-103777964 on Build GRCh38
6071164 c.1488C>T p.F496F Chr.12: 103777988-103777988 on Build GRCh38
2064311 c.1434C>T p.S478S Chr.12: 103778042-103778042 on Build GRCh38
1146889 c.1424A>G p.Q475R Chr.12: 103778052-103778052 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs759358869 Chr.12: 103778003-103778003 on Build GRCh38
rs762014599 Chr.12: 103778022-103778022 on Build GRCh38
rs200385611 Chr.12: 103778091-103778091 on Build GRCh38
rs145856854 Chr.12: 103777920-103777920 on Build GRCh38
rs758814946 Chr.12: 103777943-103777943 on Build GRCh38
rs747533314 Chr.12: 103778017-103778017 on Build GRCh38
rs780042308 Chr.12: 103777993-103777999 on Build GRCh38
rs768552682 Chr.12: 103778047-103778048 on Build GRCh38
rs771399204 Chr.12: 103778051-103778051 on Build GRCh38
rs963781447 Chr.12: 103778035-103778035 on Build GRCh38
rs781695663 Chr.12: 103778064-103778064 on Build GRCh38
rs143557037 Chr.12: 103777946-103777946 on Build GRCh38
rs770761097 Chr.12: 103777993-103777993 on Build GRCh38
rs757389915 Chr.12: 103777934-103777934 on Build GRCh38
rs1053882183 Chr.12: 103777968-103777968 on Build GRCh38
rs774079019 Chr.12: 103777995-103777995 on Build GRCh38
rs747558597 Chr.12: 103777955-103777955 on Build GRCh38
rs957497652 Chr.12: 103777947-103777947 on Build GRCh38
rs137974375 Chr.12: 103778012-103778012 on Build GRCh38
rs746913835 Chr.12: 103778022-103778023 on Build GRCh38
rs145894472 Chr.12: 103778069-103778069 on Build GRCh38
rs766620816 Chr.12: 103778045-103778045 on Build GRCh38
rs750672309 Chr.12: 103778034-103778034 on Build GRCh38
rs17034524 Chr.12: 103777964-103777964 on Build GRCh38
rs900312527 Chr.12: 103778067-103778067 on Build GRCh38
rs1020141660 Chr.12: 103777971-103777971 on Build GRCh38
rs781657942 Chr.12: 103777970-103777970 on Build GRCh38
rs555284575 Chr.12: 103777988-103777988 on Build GRCh38
rs767554232 Chr.12: 103777921-103777921 on Build GRCh38
rs190319876 Chr.12: 103778055-103778055 on Build GRCh38
rs572246923 Chr.12: 103777992-103777992 on Build GRCh38
rs372156797 Chr.12: 103777925-103777925 on Build GRCh38
rs765358062 Chr.12: 103778029-103778029 on Build GRCh38
rs200767349 Chr.12: 103777987-103777987 on Build GRCh38
rs367707322 Chr.12: 103777994-103777994 on Build GRCh38
rs769267544 Chr.12: 103777982-103777982 on Build GRCh38
rs779316333 Chr.12: 103777941-103777941 on Build GRCh38
rs1045752129 Chr.12: 103777924-103777924 on Build GRCh38
rs758483066 Chr.12: 103778043-103778043 on Build GRCh38
rs747708087 Chr.12: 103777972-103777972 on Build GRCh38
rs755315974 Chr.12: 103778058-103778058 on Build GRCh38
rs750569608 Chr.12: 103777942-103777942 on Build GRCh38

View Full Product Details