Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 73998275-73998518 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
VCL
Entrez Gene ID
7414
Gene Name
vinculin
Gene Aliases
CMD1W, CMH15, HEL114, MV, MVCL
UniGene
Hs.643896
Gene Chromosome Location
Chr.10: 73998114-74120160 on Build GRCh38
Transcripts
NM_003373.3 NM_014000.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5988649 c.156C>A p.S52R Chr.10: 73998363-73998363 on Build GRCh38
4564136 c.100_101GG>AA p.G34N Chr.10: 73998307-73998308 on Build GRCh38
6345210 c.96G>T p.V32V Chr.10: 73998303-73998303 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs918271462 Chr.10: 73998477-73998477 on Build GRCh38
rs144080529 Chr.10: 73998327-73998327 on Build GRCh38
rs1021721927 Chr.10: 73998428-73998428 on Build GRCh38
rs554116863 Chr.10: 73998337-73998337 on Build GRCh38
rs568175141 Chr.10: 73998306-73998306 on Build GRCh38
rs1029592759 Chr.10: 73998393-73998404 on Build GRCh38
rs968868225 Chr.10: 73998430-73998430 on Build GRCh38
rs1031820588 Chr.10: 73998333-73998333 on Build GRCh38
rs992494505 Chr.10: 73998468-73998468 on Build GRCh38
rs139329923 Chr.10: 73998310-73998310 on Build GRCh38
rs71535778 Chr.10: 73998430-73998431 on Build GRCh38
rs375446925 Chr.10: 73998455-73998455 on Build GRCh38
rs751938777 Chr.10: 73998365-73998365 on Build GRCh38
rs901296190 Chr.10: 73998481-73998481 on Build GRCh38
rs992545879 Chr.10: 73998343-73998343 on Build GRCh38
rs960602763 Chr.10: 73998461-73998461 on Build GRCh38
rs886047216 Chr.10: 73998362-73998362 on Build GRCh38
rs780959726 Chr.10: 73998382-73998382 on Build GRCh38
rs1002156656 Chr.10: 73998454-73998454 on Build GRCh38
rs940052176 Chr.10: 73998464-73998464 on Build GRCh38
rs983257436 Chr.10: 73998453-73998453 on Build GRCh38
rs750838330 Chr.10: 73998321-73998321 on Build GRCh38
rs997516030 Chr.10: 73998384-73998384 on Build GRCh38
rs866262635 Chr.10: 73998328-73998328 on Build GRCh38
rs1039626542 Chr.10: 73998341-73998341 on Build GRCh38
rs192805804 Chr.10: 73998452-73998452 on Build GRCh38
rs932625177 Chr.10: 73998482-73998482 on Build GRCh38
rs1010293203 Chr.10: 73998427-73998427 on Build GRCh38
rs755441334 Chr.10: 73998370-73998370 on Build GRCh38
rs891860740 Chr.10: 73998422-73998422 on Build GRCh38
rs368934323 Chr.10: 73998395-73998395 on Build GRCh38

View Full Product Details