Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 124795132-124795389 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SCARB1
Entrez Gene ID
949
Gene Name
scavenger receptor class B member 1
Gene Aliases
CD36L1, CLA-1, CLA1, HDLQTL6, SR-BI, SRB1
UniGene
Hs.731377
Gene Chromosome Location
Chr.12: 124777628-124863973 on Build GRCh38
Transcripts
NM_001082959.1 NM_005505.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6884365 c.1146G>A p.M382I Chr.12: 124795251-124795251 on Build GRCh38
4858164 c.1085G>T p.G362V Chr.12: 124795201-124795201 on Build GRCh38
4858163 c.1196G>T p.G399V Chr.12: 124795201-124795201 on Build GRCh38
6884368 c.1035G>A p.M345I Chr.12: 124795251-124795251 on Build GRCh38
294796 c.1168C>T p.L390L Chr.12: 124795229-124795229 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs776458616 Chr.12: 124795276-124795279 on Build GRCh38
rs376034642 Chr.12: 124795160-124795160 on Build GRCh38
rs761363556 Chr.12: 124795310-124795310 on Build GRCh38
rs764640671 Chr.12: 124795306-124795306 on Build GRCh38
rs772192914 Chr.12: 124795268-124795268 on Build GRCh38
rs143864257 Chr.12: 124795186-124795186 on Build GRCh38
rs372759560 Chr.12: 124795323-124795323 on Build GRCh38
rs554484840 Chr.12: 124795278-124795278 on Build GRCh38
rs922513549 Chr.12: 124795370-124795370 on Build GRCh38
rs993704900 Chr.12: 124795263-124795263 on Build GRCh38
rs748231262 Chr.12: 124795264-124795264 on Build GRCh38
rs751100225 Chr.12: 124795199-124795199 on Build GRCh38
rs763451952 Chr.12: 124795196-124795196 on Build GRCh38
rs888828592 Chr.12: 124795185-124795185 on Build GRCh38
rs373880743 Chr.12: 124795296-124795296 on Build GRCh38
rs757101173 Chr.12: 124795205-124795205 on Build GRCh38
rs773915446 Chr.12: 124795297-124795297 on Build GRCh38
rs369746779 Chr.12: 124795293-124795293 on Build GRCh38
rs147238482 Chr.12: 124795362-124795362 on Build GRCh38
rs767684123 Chr.12: 124795272-124795273 on Build GRCh38
rs750434304 Chr.12: 124795215-124795215 on Build GRCh38
rs557777297 Chr.12: 124795368-124795368 on Build GRCh38
rs773752984 Chr.12: 124795168-124795168 on Build GRCh38
rs752015118 Chr.12: 124795360-124795360 on Build GRCh38
rs773699924 Chr.12: 124795272-124795272 on Build GRCh38
rs202048724 Chr.12: 124795177-124795177 on Build GRCh38
rs755252077 Chr.12: 124795254-124795254 on Build GRCh38
rs374020615 Chr.12: 124795273-124795273 on Build GRCh38
rs369260422 Chr.12: 124795229-124795229 on Build GRCh38
rs576214655 Chr.12: 124795369-124795369 on Build GRCh38
rs111680961 Chr.12: 124795247-124795247 on Build GRCh38
rs545828803 Chr.12: 124795363-124795363 on Build GRCh38
rs10396210 Chr.12: 124795275-124795275 on Build GRCh38
rs777065351 Chr.12: 124795291-124795291 on Build GRCh38
rs1010884296 Chr.12: 124795355-124795355 on Build GRCh38
rs115774142 Chr.12: 124795206-124795206 on Build GRCh38
rs749414661 Chr.12: 124795252-124795252 on Build GRCh38
rs755969350 Chr.12: 124795223-124795223 on Build GRCh38
rs192905220 Chr.12: 124795266-124795266 on Build GRCh38
rs752824183 Chr.12: 124795235-124795235 on Build GRCh38
rs368794327 Chr.12: 124795322-124795322 on Build GRCh38
rs199829257 Chr.12: 124795239-124795239 on Build GRCh38
rs761009457 Chr.12: 124795172-124795172 on Build GRCh38
rs778820807 Chr.12: 124795256-124795256 on Build GRCh38
rs889549937 Chr.12: 124795179-124795179 on Build GRCh38

View Full Product Details