Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 73451987-73452222 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MB21D1
Entrez Gene ID
115004
Gene Name
Mab-21 domain containing 1
Gene Aliases
C6orf150, cGAS, h-cGAS
UniGene
Hs.658405
Gene Chromosome Location
Chr.6: 73423711-73452330 on Build GRCh38
Transcripts
NM_138441.2 XM_017010232.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6050989 c.159C>T p.P53P Chr.6: 73452023-73452023 on Build GRCh38
6027254 c.157C>T p.P53S Chr.6: 73452025-73452025 on Build GRCh38
4925171 c.16G>A p.G6R Chr.6: 73452166-73452166 on Build GRCh38
3875429 c.33A>G p.R11R Chr.6: 73452149-73452149 on Build GRCh38
5556672 c.15C>T p.H5H Chr.6: 73452167-73452167 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs372417314 Chr.6: 73452109-73452109 on Build GRCh38
rs762757889 Chr.6: 73452088-73452088 on Build GRCh38
rs752104616 Chr.6: 73452054-73452054 on Build GRCh38
rs759786250 Chr.6: 73452179-73452179 on Build GRCh38
rs769428394 Chr.6: 73452146-73452146 on Build GRCh38
rs770727137 Chr.6: 73452169-73452169 on Build GRCh38
rs761478616 Chr.6: 73452032-73452032 on Build GRCh38
rs771350644 Chr.6: 73452089-73452089 on Build GRCh38
rs1037410849 Chr.6: 73452199-73452199 on Build GRCh38
rs768147921 Chr.6: 73452019-73452019 on Build GRCh38
rs751343807 Chr.6: 73452136-73452136 on Build GRCh38
rs760649119 Chr.6: 73452112-73452112 on Build GRCh38
rs755440893 Chr.6: 73452055-73452055 on Build GRCh38
rs774733961 Chr.6: 73452174-73452174 on Build GRCh38
rs766912294 Chr.6: 73452040-73452040 on Build GRCh38
rs757485304 Chr.6: 73452131-73452131 on Build GRCh38
rs147035222 Chr.6: 73452132-73452132 on Build GRCh38
rs777938732 Chr.6: 73452067-73452067 on Build GRCh38
rs34985291 Chr.6: 73452017-73452017 on Build GRCh38
rs34605178 Chr.6: 73452157-73452157 on Build GRCh38
rs966509155 Chr.6: 73452117-73452117 on Build GRCh38
rs774876823 Chr.6: 73452158-73452158 on Build GRCh38
rs777423092 Chr.6: 73452148-73452148 on Build GRCh38
rs752738906 Chr.6: 73452062-73452062 on Build GRCh38
rs761236333 Chr.6: 73452195-73452195 on Build GRCh38
rs901219481 Chr.6: 73452058-73452058 on Build GRCh38
rs746906830 Chr.6: 73452072-73452072 on Build GRCh38
rs754108240 Chr.6: 73452068-73452068 on Build GRCh38
rs779830325 Chr.6: 73452070-73452070 on Build GRCh38
rs1024507349 Chr.6: 73452050-73452050 on Build GRCh38
rs373998751 Chr.6: 73452038-73452038 on Build GRCh38
rs148933122 Chr.6: 73452137-73452137 on Build GRCh38
rs768502066 Chr.6: 73452077-73452077 on Build GRCh38
rs769481836 Chr.6: 73452082-73452082 on Build GRCh38
rs774867546 Chr.6: 73452100-73452100 on Build GRCh38
rs772534763 Chr.6: 73452188-73452188 on Build GRCh38
rs775846900 Chr.6: 73452194-73452194 on Build GRCh38
rs367698486 Chr.6: 73452113-73452113 on Build GRCh38
rs35629782 Chr.6: 73452039-73452039 on Build GRCh38
rs943144752 Chr.6: 73452173-73452173 on Build GRCh38
rs888730618 Chr.6: 73452026-73452026 on Build GRCh38
rs9352000 Chr.6: 73452078-73452078 on Build GRCh38
rs748178279 Chr.6: 73452143-73452143 on Build GRCh38
rs769976348 Chr.6: 73452022-73452022 on Build GRCh38
rs767939944 Chr.6: 73452108-73452108 on Build GRCh38
rs767573211 Chr.6: 73452061-73452061 on Build GRCh38
rs758174938 Chr.6: 73452069-73452069 on Build GRCh38
rs753993852 Chr.6: 73452120-73452120 on Build GRCh38
rs539927816 Chr.6: 73452014-73452014 on Build GRCh38
rs754846031 Chr.6: 73452139-73452139 on Build GRCh38
rs540024740 Chr.6: 73452047-73452047 on Build GRCh38
rs772990516 Chr.6: 73452084-73452084 on Build GRCh38
rs756318362 Chr.6: 73452063-73452063 on Build GRCh38
rs530437247 Chr.6: 73452140-73452140 on Build GRCh38
rs760036881 Chr.6: 73452107-73452107 on Build GRCh38
rs926391717 Chr.6: 73452144-73452144 on Build GRCh38
rs1048612855 Chr.6: 73452025-73452025 on Build GRCh38

View Full Product Details