Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 101634459-101634720 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MYBPC1
Entrez Gene ID
4604
Gene Name
myosin binding protein C, slow type
Gene Aliases
LCCS4, MYBPCC, MYBPCS
UniGene
Hs.654589
Gene Chromosome Location
Chr.12: 101594849-101695841 on Build GRCh38
Transcripts
NM_001254718.1 NM_001254719.1 NM_001254720.1 NM_001254721.1 NM_001254722.1 NM_001254723.1 NM_002465.3 NM_206819.2 NM_206820.2 NM_206821.2 XM_005268876.4 XM_006719405.3 XM_006719406.3 XM_006719407.3 XM_006719408.3 XM_006719409.3 XM_006719410.3 XM_006719411.3 XM_017019315.1 XM_017019316.1 XM_017019317.1 XM_017019318.1 XM_017019319.1 XM_017019320.1 XM_017019321.1 XM_017019322.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5099057 c.482-1G>T p.? Chr.12: 101634553-101634553 on Build GRCh38
340048 c.575C>G p.P192R Chr.12: 101634572-101634572 on Build GRCh38
4979695 c.498T>C p.T166T Chr.12: 101634570-101634570 on Build GRCh38
5569324 c.489T>G p.T163T Chr.12: 101634561-101634561 on Build GRCh38
313006 c.533G>T p.S178I Chr.12: 101634605-101634605 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs111385804 Chr.12: 101634588-101634588 on Build GRCh38
rs545202429 Chr.12: 101634657-101634657 on Build GRCh38
rs748254931 Chr.12: 101634651-101634651 on Build GRCh38
rs776698100 Chr.12: 101634512-101634512 on Build GRCh38
rs780839371 Chr.12: 101634627-101634627 on Build GRCh38
rs75003679 Chr.12: 101634630-101634630 on Build GRCh38
rs529365953 Chr.12: 101634544-101634544 on Build GRCh38
rs948845543 Chr.12: 101634572-101634572 on Build GRCh38
rs11110900 Chr.12: 101634666-101634666 on Build GRCh38
rs371262248 Chr.12: 101634521-101634521 on Build GRCh38
rs751030279 Chr.12: 101634525-101634525 on Build GRCh38
rs111858076 Chr.12: 101634606-101634606 on Build GRCh38
rs892076294 Chr.12: 101634628-101634628 on Build GRCh38
rs183524782 Chr.12: 101634614-101634614 on Build GRCh38
rs773575400 Chr.12: 101634520-101634520 on Build GRCh38
rs368736837 Chr.12: 101634648-101634648 on Build GRCh38
rs373900994 Chr.12: 101634555-101634555 on Build GRCh38
rs1046652621 Chr.12: 101634539-101634539 on Build GRCh38
rs79442861 Chr.12: 101634591-101634591 on Build GRCh38
rs571874845 Chr.12: 101634652-101634652 on Build GRCh38
rs967144501 Chr.12: 101634650-101634650 on Build GRCh38
rs111414138 Chr.12: 101634552-101634552 on Build GRCh38
rs762156131 Chr.12: 101634513-101634513 on Build GRCh38
rs781402036 Chr.12: 101634642-101634642 on Build GRCh38
rs755692810 Chr.12: 101634691-101634691 on Build GRCh38
rs779819042 Chr.12: 101634534-101634534 on Build GRCh38
rs4448745 Chr.12: 101634619-101634619 on Build GRCh38
rs1048608555 Chr.12: 101634491-101634491 on Build GRCh38
rs753703767 Chr.12: 101634611-101634611 on Build GRCh38
rs190902032 Chr.12: 101634518-101634518 on Build GRCh38
rs768818531 Chr.12: 101634506-101634506 on Build GRCh38
rs754417764 Chr.12: 101634541-101634541 on Build GRCh38
rs113552077 Chr.12: 101634622-101634622 on Build GRCh38
rs569722801 Chr.12: 101634690-101634690 on Build GRCh38
rs780693699 Chr.12: 101634655-101634655 on Build GRCh38

View Full Product Details