Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 60172936-60173193 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MS4A6A
Entrez Gene ID
64231
Gene Name
membrane spanning 4-domains A6A
Gene Aliases
4SPAN3, 4SPAN3.2, CD20L3, CDA01, MS4A6, MST090, MSTP090
UniGene
Hs.523702
Gene Chromosome Location
Chr.11: 60171607-60184666 on Build GRCh38
Transcripts
NM_001247999.1 NM_022349.3 NM_152851.2 NM_152852.2 XM_005274177.2 XM_006718658.2 XM_006718660.2 XM_006718661.2 XM_017018123.1 XM_017018124.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7317628 c.580C>G p.L194V Chr.11: 60173099-60173099 on Build GRCh38
7332336 c.564G>A p.L188L Chr.11: 60173115-60173115 on Build GRCh38
6457519 c.693T>G p.T231T Chr.11: 60173070-60173070 on Build GRCh38
4615458 c.563_564insCTCT p.M189fs*27 Chr.11: 60173115-60173116 on Build GRCh38
4556923 c.623G>A p.W208* Chr.11: 60173056-60173056 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs903229733 Chr.11: 60172968-60172968 on Build GRCh38
rs751305561 Chr.11: 60172981-60172981 on Build GRCh38
rs750953546 Chr.11: 60173079-60173079 on Build GRCh38
rs752214934 Chr.11: 60173075-60173075 on Build GRCh38
rs900570842 Chr.11: 60173119-60173119 on Build GRCh38
rs762785825 Chr.11: 60173056-60173056 on Build GRCh38
rs770904421 Chr.11: 60173136-60173136 on Build GRCh38
rs752448037 Chr.11: 60173152-60173152 on Build GRCh38
rs7232 Chr.11: 60173126-60173126 on Build GRCh38
rs754812930 Chr.11: 60173149-60173149 on Build GRCh38
rs779319959 Chr.11: 60173006-60173006 on Build GRCh38
rs949128115 Chr.11: 60172974-60172974 on Build GRCh38
rs200473614 Chr.11: 60173034-60173034 on Build GRCh38
rs780314709 Chr.11: 60173023-60173023 on Build GRCh38
rs750049564 Chr.11: 60173010-60173010 on Build GRCh38
rs750179389 Chr.11: 60172961-60172961 on Build GRCh38
rs780914929 Chr.11: 60172985-60172985 on Build GRCh38
rs374502941 Chr.11: 60173000-60173000 on Build GRCh38
rs371493203 Chr.11: 60173060-60173060 on Build GRCh38
rs748846989 Chr.11: 60173165-60173165 on Build GRCh38
rs768815648 Chr.11: 60173103-60173103 on Build GRCh38
rs17852114 Chr.11: 60173124-60173124 on Build GRCh38
rs981883309 Chr.11: 60172992-60172992 on Build GRCh38
rs772374894 Chr.11: 60173015-60173015 on Build GRCh38
rs779562305 Chr.11: 60173131-60173132 on Build GRCh38
rs191121860 Chr.11: 60173134-60173134 on Build GRCh38
rs115850805 Chr.11: 60173150-60173150 on Build GRCh38
rs201213606 Chr.11: 60173078-60173078 on Build GRCh38
rs948033677 Chr.11: 60173124-60173125 on Build GRCh38
rs758155458 Chr.11: 60172978-60172978 on Build GRCh38
rs781420488 Chr.11: 60173117-60173117 on Build GRCh38
rs769727174 Chr.11: 60173044-60173044 on Build GRCh38
rs375516732 Chr.11: 60173162-60173162 on Build GRCh38
rs146398167 Chr.11: 60173059-60173059 on Build GRCh38
rs780333672 Chr.11: 60173100-60173100 on Build GRCh38
rs773187929 Chr.11: 60173132-60173132 on Build GRCh38
rs376351848 Chr.11: 60173168-60173168 on Build GRCh38
rs947357135 Chr.11: 60173064-60173064 on Build GRCh38
rs367715554 Chr.11: 60173012-60173012 on Build GRCh38
rs747384914 Chr.11: 60173102-60173102 on Build GRCh38
rs376890624 Chr.11: 60173121-60173121 on Build GRCh38
rs760222721 Chr.11: 60173157-60173157 on Build GRCh38
rs747676272 Chr.11: 60172995-60172995 on Build GRCh38
rs139777263 Chr.11: 60173062-60173062 on Build GRCh38
rs138650483 Chr.11: 60173027-60173027 on Build GRCh38
rs563501461 Chr.11: 60172979-60172979 on Build GRCh38
rs768873281 Chr.11: 60173032-60173032 on Build GRCh38
rs149285040 Chr.11: 60173140-60173140 on Build GRCh38
rs755725980 Chr.11: 60173027-60173027 on Build GRCh38
rs767302691 Chr.11: 60173071-60173071 on Build GRCh38
rs370222713 Chr.11: 60173165-60173165 on Build GRCh38
rs142963878 Chr.11: 60173091-60173091 on Build GRCh38
rs754520696 Chr.11: 60172984-60172984 on Build GRCh38

View Full Product Details