Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 57524681-57524952 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MBD6
Entrez Gene ID
114785
Gene Name
methyl-CpG binding domain protein 6
Gene Aliases
-
UniGene
Hs.524523
Gene Chromosome Location
Chr.12: 57520710-57530148 on Build GRCh38
Transcripts
NM_052897.3 XM_005268617.1 XM_005268618.1 XM_005268620.2 XM_006719217.1 XM_006719218.1 XM_011537851.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4449381 c.217-1G>A p.? Chr.12: 57524952-57524952 on Build GRCh38
6000281 c.217-10C>A p.? Chr.12: 57524943-57524943 on Build GRCh38
4776295 c.150C>A p.T50T Chr.12: 57524756-57524756 on Build GRCh38
5768176 c.114T>G p.S38R Chr.12: 57524720-57524720 on Build GRCh38
7091070 c.200C>T p.P67L Chr.12: 57524806-57524806 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs61741061 Chr.12: 57524792-57524792 on Build GRCh38
rs371781537 Chr.12: 57524706-57524706 on Build GRCh38
rs1036095325 Chr.12: 57524793-57524793 on Build GRCh38
rs752186520 Chr.12: 57524769-57524769 on Build GRCh38
rs377590759 Chr.12: 57524865-57524865 on Build GRCh38
rs757488608 Chr.12: 57524731-57524731 on Build GRCh38
rs773820030 Chr.12: 57524715-57524715 on Build GRCh38
rs763321312 Chr.12: 57524716-57524716 on Build GRCh38
rs1016747473 Chr.12: 57524729-57524729 on Build GRCh38
rs981260303 Chr.12: 57524892-57524892 on Build GRCh38
rs372878902 Chr.12: 57524908-57524908 on Build GRCh38
rs781358520 Chr.12: 57524809-57524810 on Build GRCh38
rs183250746 Chr.12: 57524713-57524713 on Build GRCh38
rs774982049 Chr.12: 57524868-57524868 on Build GRCh38
rs753532840 Chr.12: 57524801-57524801 on Build GRCh38
rs750805821 Chr.12: 57524840-57524840 on Build GRCh38
rs537746125 Chr.12: 57524818-57524818 on Build GRCh38
rs765087513 Chr.12: 57524918-57524918 on Build GRCh38
rs75754701 Chr.12: 57524877-57524877 on Build GRCh38
rs767084484 Chr.12: 57524721-57524721 on Build GRCh38
rs574510175 Chr.12: 57524893-57524893 on Build GRCh38
rs763665202 Chr.12: 57524870-57524870 on Build GRCh38
rs761321133 Chr.12: 57524912-57524912 on Build GRCh38
rs781433149 Chr.12: 57524856-57524856 on Build GRCh38
rs760417389 Chr.12: 57524779-57524779 on Build GRCh38
rs766668320 Chr.12: 57524928-57524928 on Build GRCh38
rs370728727 Chr.12: 57524907-57524907 on Build GRCh38
rs144482207 Chr.12: 57524864-57524864 on Build GRCh38
rs375118189 Chr.12: 57524929-57524929 on Build GRCh38
rs755251614 Chr.12: 57524791-57524793 on Build GRCh38
rs758206307 Chr.12: 57524825-57524825 on Build GRCh38
rs373449657 Chr.12: 57524863-57524863 on Build GRCh38
rs1148555 Chr.12: 57524851-57524851 on Build GRCh38
rs942506100 Chr.12: 57524924-57524924 on Build GRCh38
rs113877881 Chr.12: 57524885-57524885 on Build GRCh38
rs369662122 Chr.12: 57524861-57524861 on Build GRCh38
rs764915180 Chr.12: 57524814-57524814 on Build GRCh38
rs750128798 Chr.12: 57524926-57524926 on Build GRCh38
rs552864856 Chr.12: 57524845-57524845 on Build GRCh38
rs998335282 Chr.12: 57524836-57524836 on Build GRCh38
rs781571009 Chr.12: 57524841-57524841 on Build GRCh38
rs748261213 Chr.12: 57524812-57524813 on Build GRCh38
rs137897821 Chr.12: 57524808-57524808 on Build GRCh38
rs375027961 Chr.12: 57524707-57524707 on Build GRCh38

View Full Product Details