Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 9264739-9264986 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
H6PD
Entrez Gene ID
9563
Gene Name
hexose-6-phosphate dehydrogenase/glucose 1-dehydrogenase
Gene Aliases
CORTRD1, G6PDH, GDH
UniGene
Hs.463511
Gene Chromosome Location
Chr.1: 9234767-9271337 on Build GRCh38
Transcripts
NM_001282587.1 NM_004285.3 XM_005263540.4 XM_006711052.3 XM_017002865.1 XM_017002866.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1667123 c.2362G>A p.A788T Chr.1: 9264855-9264855 on Build GRCh38
4749732 c.2297A>C p.K766T Chr.1: 9264790-9264790 on Build GRCh38
4689433 c.2325G>A p.P775P Chr.1: 9264818-9264818 on Build GRCh38
6305964 c.2375G>T p.*792L Chr.1: 9264868-9264868 on Build GRCh38
3985304 c.2266A>G p.T756A Chr.1: 9264759-9264759 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs41280760 Chr.1: 9264942-9264942 on Build GRCh38
rs749654021 Chr.1: 9264895-9264895 on Build GRCh38
rs571710860 Chr.1: 9264917-9264917 on Build GRCh38
rs1013273365 Chr.1: 9264861-9264861 on Build GRCh38
rs200536036 Chr.1: 9264896-9264896 on Build GRCh38
rs372401423 Chr.1: 9264890-9264890 on Build GRCh38
rs144766765 Chr.1: 9264761-9264761 on Build GRCh38
rs531681724 Chr.1: 9264851-9264851 on Build GRCh38
rs754801418 Chr.1: 9264857-9264857 on Build GRCh38
rs766298635 Chr.1: 9264914-9264914 on Build GRCh38
rs1023159105 Chr.1: 9264803-9264803 on Build GRCh38
rs761360706 Chr.1: 9264834-9264834 on Build GRCh38
rs757440980 Chr.1: 9264874-9264874 on Build GRCh38
rs967787005 Chr.1: 9264937-9264937 on Build GRCh38
rs755938202 Chr.1: 9264852-9264852 on Build GRCh38
rs758495315 Chr.1: 9264771-9264771 on Build GRCh38
rs772652059 Chr.1: 9264908-9264908 on Build GRCh38
rs760222947 Chr.1: 9264913-9264913 on Build GRCh38
rs772793248 Chr.1: 9264825-9264825 on Build GRCh38
rs774016346 Chr.1: 9264899-9264899 on Build GRCh38
rs201296526 Chr.1: 9264760-9264760 on Build GRCh38
rs763720569 Chr.1: 9264826-9264826 on Build GRCh38
rs766210010 Chr.1: 9264854-9264854 on Build GRCh38
rs145080671 Chr.1: 9264817-9264817 on Build GRCh38
rs368809830 Chr.1: 9264778-9264778 on Build GRCh38
rs868404072 Chr.1: 9264930-9264930 on Build GRCh38
rs561393382 Chr.1: 9264824-9264824 on Build GRCh38
rs765081921 Chr.1: 9264919-9264919 on Build GRCh38
rs767020028 Chr.1: 9264905-9264905 on Build GRCh38
rs781352118 Chr.1: 9264878-9264878 on Build GRCh38
rs778183849 Chr.1: 9264863-9264863 on Build GRCh38
rs547734067 Chr.1: 9264962-9264962 on Build GRCh38
rs376390011 Chr.1: 9264891-9264891 on Build GRCh38
rs746333928 Chr.1: 9264818-9264818 on Build GRCh38
rs907965176 Chr.1: 9264815-9264815 on Build GRCh38
rs761417542 Chr.1: 9264900-9264900 on Build GRCh38
rs537164994 Chr.1: 9264931-9264931 on Build GRCh38
rs1033156144 Chr.1: 9264950-9264950 on Build GRCh38
rs753638476 Chr.1: 9264855-9264855 on Build GRCh38
rs1023795129 Chr.1: 9264933-9264933 on Build GRCh38
rs766659227 Chr.1: 9264842-9264842 on Build GRCh38
rs751301094 Chr.1: 9264957-9264957 on Build GRCh38
rs759391454 Chr.1: 9264951-9264952 on Build GRCh38
rs771105430 Chr.1: 9264806-9264806 on Build GRCh38
rs748407026 Chr.1: 9264762-9264762 on Build GRCh38
rs747214939 Chr.1: 9264873-9264873 on Build GRCh38
rs966845420 Chr.1: 9264881-9264881 on Build GRCh38
rs773959841 Chr.1: 9264829-9264829 on Build GRCh38
rs867916720 Chr.1: 9264860-9264860 on Build GRCh38
rs766967139 Chr.1: 9264843-9264843 on Build GRCh38
rs1055083445 Chr.1: 9264848-9264848 on Build GRCh38
rs1037821974 Chr.1: 9264936-9264936 on Build GRCh38
rs746395721 Chr.1: 9264885-9264885 on Build GRCh38
rs759302046 Chr.1: 9264918-9264918 on Build GRCh38
rs776563299 Chr.1: 9264855-9264855 on Build GRCh38
rs933325977 Chr.1: 9264772-9264772 on Build GRCh38
rs747155644 Chr.1: 9264797-9264797 on Build GRCh38
rs201032222 Chr.1: 9264823-9264823 on Build GRCh38

View Full Product Details