Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 52063692-52063949 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MAPK6
Entrez Gene ID
5597
Gene Name
mitogen-activated protein kinase 6
Gene Aliases
ERK3, HsT17250, PRKM6, p97MAPK
UniGene
Hs.411847
Gene Chromosome Location
Chr.15: 52004265-52066265 on Build GRCh38
Transcripts
NM_002748.3 XM_005254537.2 XM_005254538.2 XM_005254539.3 XM_011521782.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5940531 c.1088C>T p.S363L Chr.15: 52063922-52063922 on Build GRCh38
4055541 c.1102C>T p.P368S Chr.15: 52063936-52063936 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs764304859 Chr.15: 52063915-52063915 on Build GRCh38
rs35691641 Chr.15: 52063753-52063754 on Build GRCh38
rs541591673 Chr.15: 52063860-52063860 on Build GRCh38
rs762189147 Chr.15: 52063919-52063919 on Build GRCh38
rs919620905 Chr.15: 52063786-52063786 on Build GRCh38
rs759100590 Chr.15: 52063917-52063917 on Build GRCh38
rs879011408 Chr.15: 52063822-52063822 on Build GRCh38
rs940264204 Chr.15: 52063726-52063726 on Build GRCh38
rs767091620 Chr.15: 52063925-52063925 on Build GRCh38
rs769660207 Chr.15: 52063880-52063880 on Build GRCh38
rs950130549 Chr.15: 52063760-52063760 on Build GRCh38
rs930993704 Chr.15: 52063788-52063788 on Build GRCh38
rs927014489 Chr.15: 52063808-52063808 on Build GRCh38
rs745523178 Chr.15: 52063771-52063771 on Build GRCh38
rs954201263 Chr.15: 52063744-52063744 on Build GRCh38
rs199741074 Chr.15: 52063917-52063917 on Build GRCh38
rs1028371872 Chr.15: 52063738-52063738 on Build GRCh38
rs765547248 Chr.15: 52063921-52063921 on Build GRCh38
rs938534233 Chr.15: 52063812-52063812 on Build GRCh38
rs891610935 Chr.15: 52063853-52063853 on Build GRCh38
rs905929173 Chr.15: 52063764-52063764 on Build GRCh38
rs368865611 Chr.15: 52063842-52063842 on Build GRCh38
rs763413185 Chr.15: 52063863-52063873 on Build GRCh38
rs779407766 Chr.15: 52063873-52063873 on Build GRCh38
rs370355033 Chr.15: 52063884-52063884 on Build GRCh38
rs180742711 Chr.15: 52063908-52063908 on Build GRCh38
rs986937501 Chr.15: 52063749-52063749 on Build GRCh38
rs758642801 Chr.15: 52063891-52063892 on Build GRCh38
rs762771625 Chr.15: 52063906-52063906 on Build GRCh38
rs754198242 Chr.15: 52063855-52063855 on Build GRCh38
rs1037210568 Chr.15: 52063737-52063737 on Build GRCh38
rs761028406 Chr.15: 52063911-52063911 on Build GRCh38
rs188220127 Chr.15: 52063794-52063794 on Build GRCh38
rs200318726 Chr.15: 52063879-52063879 on Build GRCh38
rs972997889 Chr.15: 52063783-52063783 on Build GRCh38
rs746313015 Chr.15: 52063868-52063868 on Build GRCh38
rs377355684 Chr.15: 52063869-52063869 on Build GRCh38
rs574337540 Chr.15: 52063854-52063854 on Build GRCh38
rs778481703 Chr.15: 52063762-52063762 on Build GRCh38
rs961954352 Chr.15: 52063774-52063774 on Build GRCh38
rs766941779 Chr.15: 52063872-52063873 on Build GRCh38
rs779407583 Chr.15: 52063865-52063865 on Build GRCh38
rs979825162 Chr.15: 52063791-52063791 on Build GRCh38
rs878943357 Chr.15: 52063897-52063897 on Build GRCh38
rs1000233401 Chr.15: 52063818-52063818 on Build GRCh38
rs775423389 Chr.15: 52063909-52063909 on Build GRCh38
rs556999957 Chr.15: 52063878-52063878 on Build GRCh38

View Full Product Details