Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 45394489-45394722 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LARS2
Entrez Gene ID
23395
Gene Name
leucyl-tRNA synthetase 2, mitochondrial
Gene Aliases
LEURS, PRLTS4, mtLeuRS
UniGene
Hs.526975
Gene Chromosome Location
Chr.3: 45388561-45548842 on Build GRCh38
Transcripts
NM_015340.3 XM_005265006.1 XM_011533554.2 XM_017006042.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5638249 c.213C>T p.A71A Chr.3: 45394666-45394666 on Build GRCh38
4880884 c.38C>G p.S13C Chr.3: 45394491-45394491 on Build GRCh38
1692671 c.217A>G p.K73E Chr.3: 45394670-45394670 on Build GRCh38
4117803 c.132G>A p.T44T Chr.3: 45394585-45394585 on Build GRCh38
1423310 c.138G>T p.K46N Chr.3: 45394591-45394591 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs532725261 Chr.3: 45394668-45394668 on Build GRCh38
rs762319941 Chr.3: 45394632-45394632 on Build GRCh38
rs754475523 Chr.3: 45394672-45394672 on Build GRCh38
rs541763978 Chr.3: 45394677-45394677 on Build GRCh38
rs777694670 Chr.3: 45394598-45394598 on Build GRCh38
rs781238503 Chr.3: 45394584-45394584 on Build GRCh38
rs576608165 Chr.3: 45394585-45394585 on Build GRCh38
rs146378764 Chr.3: 45394534-45394534 on Build GRCh38
rs751261319 Chr.3: 45394669-45394669 on Build GRCh38
rs752704499 Chr.3: 45394542-45394542 on Build GRCh38
rs553596357 Chr.3: 45394655-45394655 on Build GRCh38
rs149789249 Chr.3: 45394650-45394650 on Build GRCh38
rs143095137 Chr.3: 45394588-45394588 on Build GRCh38
rs199568924 Chr.3: 45394633-45394633 on Build GRCh38
rs1020076031 Chr.3: 45394555-45394555 on Build GRCh38
rs776107034 Chr.3: 45394585-45394585 on Build GRCh38
rs370289532 Chr.3: 45394691-45394691 on Build GRCh38
rs777055524 Chr.3: 45394625-45394625 on Build GRCh38
rs901736144 Chr.3: 45394564-45394564 on Build GRCh38
rs776961881 Chr.3: 45394533-45394533 on Build GRCh38
rs779796862 Chr.3: 45394551-45394551 on Build GRCh38
rs373693561 Chr.3: 45394577-45394577 on Build GRCh38
rs772150626 Chr.3: 45394609-45394609 on Build GRCh38
rs750723957 Chr.3: 45394661-45394661 on Build GRCh38
rs768754426 Chr.3: 45394622-45394622 on Build GRCh38
rs751011983 Chr.3: 45394576-45394576 on Build GRCh38
rs141607519 Chr.3: 45394610-45394610 on Build GRCh38
rs200135115 Chr.3: 45394601-45394601 on Build GRCh38
rs763424959 Chr.3: 45394666-45394666 on Build GRCh38
rs962130384 Chr.3: 45394692-45394692 on Build GRCh38
rs113559893 Chr.3: 45394556-45394556 on Build GRCh38
rs780865850 Chr.3: 45394673-45394673 on Build GRCh38
rs997855015 Chr.3: 45394580-45394580 on Build GRCh38

View Full Product Details