Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 84926556-84926829 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MCOLN2
Entrez Gene ID
255231
Gene Name
mucolipin 2
Gene Aliases
TRP-ML2, TRPML2
UniGene
Hs.591446
Gene Chromosome Location
Chr.1: 84925583-84997122 on Build GRCh38
Transcripts
NM_153259.3 XM_005270719.3 XM_006710552.2 XM_011541187.2 XM_011541188.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
249173 c.1665G>T p.R555S Chr.1: 84926721-84926721 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs778947977 Chr.1: 84926765-84926765 on Build GRCh38
rs745741641 Chr.1: 84926739-84926739 on Build GRCh38
rs541560554 Chr.1: 84926653-84926653 on Build GRCh38
rs772627598 Chr.1: 84926763-84926763 on Build GRCh38
rs6681025 Chr.1: 84926641-84926641 on Build GRCh38
rs1046169394 Chr.1: 84926694-84926694 on Build GRCh38
rs773590804 Chr.1: 84926772-84926772 on Build GRCh38
rs755530385 Chr.1: 84926728-84926728 on Build GRCh38
rs948812329 Chr.1: 84926633-84926633 on Build GRCh38
rs993003583 Chr.1: 84926624-84926624 on Build GRCh38
rs369721684 Chr.1: 84926757-84926757 on Build GRCh38
rs750850411 Chr.1: 84926711-84926711 on Build GRCh38
rs762271104 Chr.1: 84926693-84926693 on Build GRCh38
rs748839335 Chr.1: 84926750-84926750 on Build GRCh38
rs940466490 Chr.1: 84926630-84926630 on Build GRCh38
rs141704454 Chr.1: 84926650-84926650 on Build GRCh38
rs779768021 Chr.1: 84926743-84926743 on Build GRCh38
rs761295363 Chr.1: 84926762-84926762 on Build GRCh38
rs774857938 Chr.1: 84926659-84926659 on Build GRCh38
rs756980449 Chr.1: 84926729-84926729 on Build GRCh38
rs766692176 Chr.1: 84926721-84926721 on Build GRCh38
rs769704374 Chr.1: 84926786-84926786 on Build GRCh38
rs189859632 Chr.1: 84926759-84926759 on Build GRCh38
rs752793348 Chr.1: 84926733-84926733 on Build GRCh38
rs779087654 Chr.1: 84926729-84926729 on Build GRCh38
rs753942464 Chr.1: 84926723-84926723 on Build GRCh38
rs769405574 Chr.1: 84926741-84926741 on Build GRCh38
rs767985367 Chr.1: 84926699-84926699 on Build GRCh38
rs749164472 Chr.1: 84926705-84926706 on Build GRCh38
rs780976665 Chr.1: 84926736-84926736 on Build GRCh38
rs370159008 Chr.1: 84926645-84926645 on Build GRCh38
rs34293401 Chr.1: 84926758-84926758 on Build GRCh38
rs757308860 Chr.1: 84926735-84926735 on Build GRCh38
rs200768372 Chr.1: 84926640-84926640 on Build GRCh38
rs200005364 Chr.1: 84926717-84926717 on Build GRCh38

View Full Product Details