Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 139598295-139598568 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MCF2
Entrez Gene ID
4168
Gene Name
MCF.2 cell line derived transforming sequence
Gene Aliases
ARHGEF21, DBL
UniGene
Hs.387262
Gene Chromosome Location
Chr.X: 139581768-139708279 on Build GRCh38
Transcripts
NM_001099855.1 NM_001171876.1 NM_001171877.1 NM_001171878.1 NM_001171879.1 NM_005369.4 XM_005262413.4 XM_011531339.2 XM_017029529.1 XM_017029530.1 XM_017029531.1 XM_017029532.1

SNP Detail

SNP ID Chromosome Location
rs370241205 Chr.X: 139598434-139598434 on Build GRCh38
rs17002165 Chr.X: 139598346-139598346 on Build GRCh38
rs34201792 Chr.X: 139598378-139598379 on Build GRCh38
rs772543314 Chr.X: 139598527-139598527 on Build GRCh38
rs749468590 Chr.X: 139598399-139598399 on Build GRCh38
rs756234303 Chr.X: 139598395-139598395 on Build GRCh38
rs769352956 Chr.X: 139598350-139598350 on Build GRCh38
rs906120041 Chr.X: 139598391-139598391 on Build GRCh38
rs761209849 Chr.X: 139598545-139598545 on Build GRCh38
rs150405603 Chr.X: 139598427-139598427 on Build GRCh38
rs778344008 Chr.X: 139598525-139598529 on Build GRCh38
rs994424884 Chr.X: 139598467-139598467 on Build GRCh38
rs773685793 Chr.X: 139598532-139598532 on Build GRCh38
rs770250622 Chr.X: 139598435-139598435 on Build GRCh38
rs184864923 Chr.X: 139598424-139598424 on Build GRCh38
rs751600356 Chr.X: 139598372-139598372 on Build GRCh38
rs866947207 Chr.X: 139598499-139598499 on Build GRCh38
rs781760872 Chr.X: 139598382-139598382 on Build GRCh38

View Full Product Details