Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 48917664-48917935 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PIM2
Entrez Gene ID
11040
Gene Name
Pim-2 proto-oncogene, serine/threonine kinase
Gene Aliases
-
UniGene
Hs.727148
Gene Chromosome Location
Chr.X: 48913182-48919136 on Build GRCh38
Transcripts
NM_006875.3

SNP Detail

SNP ID Chromosome Location
rs782274472 Chr.X: 48917888-48917888 on Build GRCh38
rs980893139 Chr.X: 48917857-48917857 on Build GRCh38
rs782328453 Chr.X: 48917748-48917748 on Build GRCh38
rs782759236 Chr.X: 48917847-48917847 on Build GRCh38
rs141748644 Chr.X: 48917689-48917689 on Build GRCh38
rs782005307 Chr.X: 48917835-48917835 on Build GRCh38
rs782025967 Chr.X: 48917786-48917786 on Build GRCh38
rs781787847 Chr.X: 48917858-48917858 on Build GRCh38
rs782172388 Chr.X: 48917797-48917797 on Build GRCh38
rs782388667 Chr.X: 48917737-48917737 on Build GRCh38
rs782784384 Chr.X: 48917901-48917901 on Build GRCh38
rs782185628 Chr.X: 48917746-48917746 on Build GRCh38
rs782080022 Chr.X: 48917842-48917842 on Build GRCh38
rs782792364 Chr.X: 48917865-48917865 on Build GRCh38
rs782378597 Chr.X: 48917811-48917811 on Build GRCh38
rs782125448 Chr.X: 48917725-48917725 on Build GRCh38
rs782063587 Chr.X: 48917863-48917863 on Build GRCh38
rs781901723 Chr.X: 48917911-48917911 on Build GRCh38
rs782585508 Chr.X: 48917744-48917744 on Build GRCh38

View Full Product Details