Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 75556027-75556254 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LLGL2
Entrez Gene ID
3993
Gene Name
LLGL2, scribble cell polarity complex component
Gene Aliases
HGL, Hugl-2, LGL2
UniGene
Hs.514477
Gene Chromosome Location
Chr.17: 75525080-75575209 on Build GRCh38
Transcripts
NM_001015002.1 NM_001031803.1 NM_004524.2 XM_011524801.1 XM_011524802.1 XM_011524807.2 XM_017024625.1 XM_017024626.1 XM_017024627.1 XM_017024628.1 XM_017024629.1 XM_017024630.1 XM_017024631.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4580107 c.77C>T p.T26M Chr.17: 75556047-75556047 on Build GRCh38
6454478 c.78G>A p.T26T Chr.17: 75556048-75556048 on Build GRCh38
1134492 c.76-1G>A p.? Chr.17: 75556045-75556045 on Build GRCh38
3890402 c.93C>T p.F31F Chr.17: 75556063-75556063 on Build GRCh38
1732404 c.124C>G p.P42A Chr.17: 75556094-75556094 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs769584328 Chr.17: 75556099-75556099 on Build GRCh38
rs920473906 Chr.17: 75556056-75556056 on Build GRCh38
rs761967911 Chr.17: 75556118-75556118 on Build GRCh38
rs374538010 Chr.17: 75556152-75556152 on Build GRCh38
rs568230637 Chr.17: 75556153-75556153 on Build GRCh38
rs762917323 Chr.17: 75556124-75556124 on Build GRCh38
rs762057705 Chr.17: 75556078-75556078 on Build GRCh38
rs972037964 Chr.17: 75556168-75556168 on Build GRCh38
rs780391924 Chr.17: 75556087-75556087 on Build GRCh38
rs778516875 Chr.17: 75556167-75556167 on Build GRCh38
rs755244871 Chr.17: 75556094-75556094 on Build GRCh38
rs758567092 Chr.17: 75556084-75556084 on Build GRCh38
rs150649144 Chr.17: 75556123-75556123 on Build GRCh38
rs745841597 Chr.17: 75556115-75556115 on Build GRCh38
rs911943346 Chr.17: 75556160-75556160 on Build GRCh38
rs748801213 Chr.17: 75556103-75556103 on Build GRCh38
rs368567805 Chr.17: 75556048-75556048 on Build GRCh38
rs371012842 Chr.17: 75556135-75556135 on Build GRCh38
rs765238069 Chr.17: 75556079-75556079 on Build GRCh38
rs772853182 Chr.17: 75556054-75556054 on Build GRCh38
rs138134053 Chr.17: 75556047-75556047 on Build GRCh38
rs769387744 Chr.17: 75556051-75556051 on Build GRCh38
rs756612177 Chr.17: 75556161-75556161 on Build GRCh38
rs144195122 Chr.17: 75556117-75556117 on Build GRCh38
rs376986902 Chr.17: 75556069-75556069 on Build GRCh38
rs762924049 Chr.17: 75556063-75556063 on Build GRCh38
rs556637068 Chr.17: 75556201-75556201 on Build GRCh38
rs570259692 Chr.17: 75556228-75556228 on Build GRCh38
rs889328205 Chr.17: 75556174-75556174 on Build GRCh38
rs777624951 Chr.17: 75556100-75556100 on Build GRCh38
rs552446966 Chr.17: 75556067-75556067 on Build GRCh38
rs142615248 Chr.17: 75556095-75556095 on Build GRCh38
rs146029134 Chr.17: 75556096-75556096 on Build GRCh38
rs774136253 Chr.17: 75556108-75556108 on Build GRCh38
rs1671036 Chr.17: 75556104-75556104 on Build GRCh38
rs905053501 Chr.17: 75556085-75556085 on Build GRCh38
rs766498095 Chr.17: 75556125-75556125 on Build GRCh38
rs766641401 Chr.17: 75556055-75556055 on Build GRCh38
rs930542197 Chr.17: 75556209-75556209 on Build GRCh38
rs201480495 Chr.17: 75556188-75556188 on Build GRCh38
rs752154844 Chr.17: 75556090-75556090 on Build GRCh38
rs372327675 Chr.17: 75556066-75556066 on Build GRCh38
rs755544035 Chr.17: 75556065-75556065 on Build GRCh38
rs756673867 Chr.17: 75556164-75556164 on Build GRCh38
rs988061264 Chr.17: 75556157-75556157 on Build GRCh38
rs1008206292 Chr.17: 75556224-75556224 on Build GRCh38
rs868300040 Chr.17: 75556091-75556091 on Build GRCh38
rs201650903 Chr.17: 75556170-75556170 on Build GRCh38
rs920631201 Chr.17: 75556206-75556206 on Build GRCh38

View Full Product Details