Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 70532666-70532925 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PALD1
Entrez Gene ID
27143
Gene Name
phosphatase domain containing, paladin 1
Gene Aliases
KIAA1274, PALD
UniGene
Hs.202351
Gene Chromosome Location
Chr.10: 70458542-70568450 on Build GRCh38
Transcripts
NM_014431.2 XM_011539637.2 XM_011539638.2 XM_017016072.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7104734 c.748A>G p.T250A Chr.10: 70532735-70532735 on Build GRCh38
5812209 c.744T>A p.H248Q Chr.10: 70532731-70532731 on Build GRCh38
1348838 c.718G>A p.A240T Chr.10: 70532705-70532705 on Build GRCh38
7155698 c.723C>G p.I241M Chr.10: 70532710-70532710 on Build GRCh38
6482680 c.720C>G p.A240A Chr.10: 70532707-70532707 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs751826004 Chr.10: 70532821-70532821 on Build GRCh38
rs767927106 Chr.10: 70532830-70532830 on Build GRCh38
rs772344991 Chr.10: 70532871-70532871 on Build GRCh38
rs760602665 Chr.10: 70532804-70532804 on Build GRCh38
rs773997401 Chr.10: 70532714-70532714 on Build GRCh38
rs368529278 Chr.10: 70532891-70532891 on Build GRCh38
rs141669123 Chr.10: 70532735-70532735 on Build GRCh38
rs755614864 Chr.10: 70532768-70532768 on Build GRCh38
rs773586927 Chr.10: 70532872-70532872 on Build GRCh38
rs148437569 Chr.10: 70532722-70532722 on Build GRCh38
rs773251889 Chr.10: 70532808-70532808 on Build GRCh38
rs750570062 Chr.10: 70532745-70532745 on Build GRCh38
rs887150745 Chr.10: 70532859-70532859 on Build GRCh38
rs201606863 Chr.10: 70532788-70532788 on Build GRCh38
rs145425283 Chr.10: 70532710-70532710 on Build GRCh38
rs771868448 Chr.10: 70532728-70532728 on Build GRCh38
rs1036306065 Chr.10: 70532900-70532900 on Build GRCh38
rs977671439 Chr.10: 70532758-70532758 on Build GRCh38
rs748950169 Chr.10: 70532701-70532701 on Build GRCh38
rs755400949 Chr.10: 70532759-70532759 on Build GRCh38
rs2275061 Chr.10: 70532761-70532761 on Build GRCh38
rs376749708 Chr.10: 70532810-70532810 on Build GRCh38
rs753175944 Chr.10: 70532832-70532832 on Build GRCh38
rs748595665 Chr.10: 70532767-70532767 on Build GRCh38
rs571686428 Chr.10: 70532850-70532850 on Build GRCh38
rs751838960 Chr.10: 70532755-70532755 on Build GRCh38
rs368026129 Chr.10: 70532793-70532793 on Build GRCh38
rs142191615 Chr.10: 70532689-70532689 on Build GRCh38
rs569796890 Chr.10: 70532855-70532855 on Build GRCh38
rs759757379 Chr.10: 70532828-70532828 on Build GRCh38
rs777411160 Chr.10: 70532771-70532771 on Build GRCh38
rs888682510 Chr.10: 70532811-70532811 on Build GRCh38
rs745602119 Chr.10: 70532790-70532790 on Build GRCh38
rs769447017 Chr.10: 70532686-70532686 on Build GRCh38
rs376431690 Chr.10: 70532753-70532753 on Build GRCh38
rs372414859 Chr.10: 70532785-70532785 on Build GRCh38
rs766671595 Chr.10: 70532754-70532754 on Build GRCh38
rs1014338369 Chr.10: 70532856-70532856 on Build GRCh38
rs373712496 Chr.10: 70532737-70532737 on Build GRCh38
rs191087909 Chr.10: 70532693-70532693 on Build GRCh38
rs1055862550 Chr.10: 70532792-70532792 on Build GRCh38
rs149222447 Chr.10: 70532795-70532795 on Build GRCh38
rs369552211 Chr.10: 70532736-70532736 on Build GRCh38
rs766470547 Chr.10: 70532812-70532812 on Build GRCh38
rs1032837686 Chr.10: 70532906-70532906 on Build GRCh38
rs748858432 Chr.10: 70532774-70532774 on Build GRCh38
rs967551860 Chr.10: 70532892-70532892 on Build GRCh38
rs753267660 Chr.10: 70532766-70532766 on Build GRCh38
rs1004607715 Chr.10: 70532881-70532881 on Build GRCh38

View Full Product Details