Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 93740793-93741018 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM172A
Entrez Gene ID
83989
Gene Name
family with sequence similarity 172 member A
Gene Aliases
C5orf21
UniGene
Hs.600086
Gene Chromosome Location
Chr.5: 93615130-94111699 on Build GRCh38
Transcripts
NM_001163417.1 NM_001163418.1 NM_032042.5 NR_028080.1 XM_005272105.3 XM_005272106.3 XM_005272108.3 XM_005272110.3 XM_006714717.3 XM_006714718.3 XM_006714719.3 XM_011543671.2 XM_011543672.2 XM_011543673.2 XM_017009952.1 XM_017009953.1 XM_017009955.1 XM_017009956.1 XM_017009957.1 XM_017009958.1 XM_017009960.1 NM_153216.1

Gene Symbol
POU5F2
Entrez Gene ID
134187
Gene Name
POU domain class 5, transcription factor 2
Gene Aliases
SPRM-1
UniGene
Hs.678995
Gene Chromosome Location
Chr.5: 93740309-93741603 on Build GRCh38
Transcripts
NM_001163417.1 NM_001163418.1 NM_032042.5 NR_028080.1 XM_005272105.3 XM_005272106.3 XM_005272108.3 XM_005272110.3 XM_006714717.3 XM_006714718.3 XM_006714719.3 XM_011543671.2 XM_011543672.2 XM_011543673.2 XM_017009952.1 XM_017009953.1 XM_017009955.1 XM_017009956.1 XM_017009957.1 XM_017009958.1 XM_017009960.1 NM_153216.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6753035 c.691C>T p.R231W Chr.5: 93740873-93740873 on Build GRCh38
3393715 c.598T>G p.C200G Chr.5: 93740966-93740966 on Build GRCh38
3828603 c.636G>C p.W212C Chr.5: 93740928-93740928 on Build GRCh38
5809023 c.669C>A p.N223K Chr.5: 93740895-93740895 on Build GRCh38
3994515 c.588T>A p.L196L Chr.5: 93740976-93740976 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs746749908 Chr.5: 93740882-93740882 on Build GRCh38
rs754303500 Chr.5: 93740829-93740829 on Build GRCh38
rs757032596 Chr.5: 93740970-93740970 on Build GRCh38
rs748177777 Chr.5: 93740993-93740993 on Build GRCh38
rs781148855 Chr.5: 93740895-93740895 on Build GRCh38
rs779169489 Chr.5: 93740902-93740902 on Build GRCh38
rs777533339 Chr.5: 93740876-93740876 on Build GRCh38
rs554717601 Chr.5: 93740949-93740949 on Build GRCh38
rs748374007 Chr.5: 93740904-93740904 on Build GRCh38
rs750340501 Chr.5: 93740983-93740983 on Build GRCh38
rs543468590 Chr.5: 93740905-93740905 on Build GRCh38
rs754470635 Chr.5: 93740845-93740845 on Build GRCh38
rs757009479 Chr.5: 93740886-93740886 on Build GRCh38
rs745818915 Chr.5: 93740900-93740900 on Build GRCh38
rs369080417 Chr.5: 93740823-93740823 on Build GRCh38
rs756069005 Chr.5: 93740984-93740984 on Build GRCh38
rs764675526 Chr.5: 93740869-93740869 on Build GRCh38
rs1042110357 Chr.5: 93740854-93740854 on Build GRCh38
rs377225323 Chr.5: 93740940-93740940 on Build GRCh38
rs552948117 Chr.5: 93740874-93740874 on Build GRCh38
rs777147043 Chr.5: 93740912-93740912 on Build GRCh38
rs752263779 Chr.5: 93740944-93740944 on Build GRCh38
rs775400558 Chr.5: 93740986-93740986 on Build GRCh38
rs771230898 Chr.5: 93740908-93740908 on Build GRCh38
rs1056965537 Chr.5: 93740917-93740917 on Build GRCh38
rs1017135332 Chr.5: 93740931-93740931 on Build GRCh38
rs938075584 Chr.5: 93740910-93740910 on Build GRCh38
rs780901290 Chr.5: 93740971-93740971 on Build GRCh38
rs762465212 Chr.5: 93740948-93740948 on Build GRCh38
rs763672762 Chr.5: 93740967-93740967 on Build GRCh38
rs199511635 Chr.5: 93740901-93740901 on Build GRCh38
rs751246695 Chr.5: 93740969-93740969 on Build GRCh38
rs371786597 Chr.5: 93740872-93740872 on Build GRCh38
rs761032270 Chr.5: 93740906-93740906 on Build GRCh38
rs760002979 Chr.5: 93740921-93740921 on Build GRCh38
rs926613223 Chr.5: 93740880-93740880 on Build GRCh38

View Full Product Details