Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 3071725-3071996 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MYOM1
Entrez Gene ID
8736
Gene Name
myomesin 1
Gene Aliases
SKELEMIN
UniGene
Hs.464469
Gene Chromosome Location
Chr.18: 3066807-3220108 on Build GRCh38
Transcripts
NM_003803.3 NM_019856.1 XM_017026062.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5972586 c.4743G>A p.V1581V Chr.18: 3071855-3071855 on Build GRCh38
5018920 c.4763delA p.K1588fs*20 Chr.18: 3071835-3071835 on Build GRCh38
7249146 c.4718G>A p.R1573Q Chr.18: 3071880-3071880 on Build GRCh38
344637 c.4764G>A p.K1588K Chr.18: 3071834-3071834 on Build GRCh38
473767 c.4709-2A>T p.? Chr.18: 3071891-3071891 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1006339081 Chr.18: 3071763-3071763 on Build GRCh38
rs559625778 Chr.18: 3071881-3071881 on Build GRCh38
rs1046921731 Chr.18: 3071965-3071965 on Build GRCh38
rs117342470 Chr.18: 3071880-3071880 on Build GRCh38
rs781184954 Chr.18: 3071837-3071837 on Build GRCh38
rs763663857 Chr.18: 3071916-3071916 on Build GRCh38
rs764033190 Chr.18: 3071805-3071805 on Build GRCh38
rs748345217 Chr.18: 3071901-3071901 on Build GRCh38
rs929903251 Chr.18: 3071954-3071954 on Build GRCh38
rs769419471 Chr.18: 3071899-3071899 on Build GRCh38
rs752316304 Chr.18: 3071795-3071795 on Build GRCh38
rs775018515 Chr.18: 3071911-3071911 on Build GRCh38
rs907072128 Chr.18: 3071755-3071755 on Build GRCh38
rs377065011 Chr.18: 3071938-3071938 on Build GRCh38
rs373419422 Chr.18: 3071887-3071887 on Build GRCh38
rs878948438 Chr.18: 3071752-3071752 on Build GRCh38
rs753268904 Chr.18: 3071759-3071759 on Build GRCh38
rs750662926 Chr.18: 3071848-3071848 on Build GRCh38
rs79745437 Chr.18: 3071824-3071824 on Build GRCh38
rs996587559 Chr.18: 3071817-3071817 on Build GRCh38
rs759105315 Chr.18: 3071898-3071898 on Build GRCh38
rs1045521955 Chr.18: 3071747-3071747 on Build GRCh38
rs955565448 Chr.18: 3071937-3071937 on Build GRCh38
rs543378017 Chr.18: 3071770-3071770 on Build GRCh38
rs888318759 Chr.18: 3071968-3071968 on Build GRCh38
rs764637678 Chr.18: 3071784-3071784 on Build GRCh38
rs751579824 Chr.18: 3071813-3071813 on Build GRCh38
rs1029829638 Chr.18: 3071910-3071910 on Build GRCh38
rs924855224 Chr.18: 3071960-3071960 on Build GRCh38
rs756428714 Chr.18: 3071855-3071855 on Build GRCh38
rs768645052 Chr.18: 3071876-3071876 on Build GRCh38
rs916319174 Chr.18: 3071896-3071896 on Build GRCh38
rs1016673799 Chr.18: 3071766-3071766 on Build GRCh38
rs369605033 Chr.18: 3071857-3071857 on Build GRCh38
rs900458971 Chr.18: 3071809-3071809 on Build GRCh38
rs369967493 Chr.18: 3071886-3071886 on Build GRCh38
rs748689826 Chr.18: 3071884-3071884 on Build GRCh38
rs781591383 Chr.18: 3071868-3071870 on Build GRCh38
rs201979586 Chr.18: 3071858-3071858 on Build GRCh38
rs762373774 Chr.18: 3071801-3071801 on Build GRCh38
rs760062519 Chr.18: 3071897-3071897 on Build GRCh38
rs375909964 Chr.18: 3071789-3071789 on Build GRCh38

View Full Product Details