Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 113705967-113706222 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PHTF1
Entrez Gene ID
10745
Gene Name
putative homeodomain transcription factor 1
Gene Aliases
PHTF
UniGene
Hs.655824
Gene Chromosome Location
Chr.1: 113697202-113759888 on Build GRCh38
Transcripts
NM_001323041.1 NM_001323042.1 NM_001323043.1 NM_001323044.1 NM_001323045.1 NM_001323046.1 NM_001323047.1 NM_001323048.1 NM_001323049.1 NM_001323050.1 NM_001323051.1 NM_001323052.1 NM_001323053.1 NM_006608.2 NR_136564.1 NR_136565.1 NR_136566.1 NR_136567.1 NR_136568.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2120298 c.1576A>G p.I526V Chr.1: 113705985-113705985 on Build GRCh38
5349943 c.1546C>G p.L516V Chr.1: 113706015-113706015 on Build GRCh38
1332808 c.1484G>A p.R495Q Chr.1: 113706077-113706077 on Build GRCh38
5843100 c.1486C>T p.L496F Chr.1: 113706075-113706075 on Build GRCh38
6745667 c.1483C>G p.R495G Chr.1: 113706078-113706078 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs753648508 Chr.1: 113706118-113706118 on Build GRCh38
rs765720779 Chr.1: 113706181-113706181 on Build GRCh38
rs752359179 Chr.1: 113706143-113706143 on Build GRCh38
rs746535381 Chr.1: 113706158-113706158 on Build GRCh38
rs140005268 Chr.1: 113706069-113706069 on Build GRCh38
rs368367033 Chr.1: 113706099-113706099 on Build GRCh38
rs894797072 Chr.1: 113706077-113706077 on Build GRCh38
rs763565407 Chr.1: 113706169-113706169 on Build GRCh38
rs764311116 Chr.1: 113706008-113706008 on Build GRCh38
rs777185000 Chr.1: 113706063-113706063 on Build GRCh38
rs754614592 Chr.1: 113706049-113706049 on Build GRCh38
rs764987109 Chr.1: 113706141-113706141 on Build GRCh38
rs770312628 Chr.1: 113706014-113706014 on Build GRCh38
rs374862815 Chr.1: 113706187-113706187 on Build GRCh38
rs774226564 Chr.1: 113706017-113706017 on Build GRCh38
rs767544967 Chr.1: 113706032-113706032 on Build GRCh38
rs867145399 Chr.1: 113706173-113706173 on Build GRCh38
rs374444021 Chr.1: 113706057-113706057 on Build GRCh38
rs754839422 Chr.1: 113706129-113706129 on Build GRCh38
rs145965579 Chr.1: 113706003-113706003 on Build GRCh38
rs541404540 Chr.1: 113706103-113706103 on Build GRCh38
rs778041552 Chr.1: 113706172-113706172 on Build GRCh38
rs763148035 Chr.1: 113706002-113706002 on Build GRCh38
rs371150761 Chr.1: 113706034-113706034 on Build GRCh38
rs749656850 Chr.1: 113706078-113706078 on Build GRCh38
rs545835977 Chr.1: 113706188-113706188 on Build GRCh38
rs79135507 Chr.1: 113706090-113706090 on Build GRCh38
rs147693102 Chr.1: 113706159-113706159 on Build GRCh38
rs766266692 Chr.1: 113706043-113706043 on Build GRCh38
rs768819745 Chr.1: 113706082-113706082 on Build GRCh38
rs140067418 Chr.1: 113706152-113706152 on Build GRCh38
rs148623704 Chr.1: 113706068-113706068 on Build GRCh38
rs1036789183 Chr.1: 113706021-113706021 on Build GRCh38
rs757898598 Chr.1: 113706059-113706059 on Build GRCh38
rs371832363 Chr.1: 113706117-113706117 on Build GRCh38
rs779213481 Chr.1: 113706195-113706195 on Build GRCh38
rs758049269 Chr.1: 113706150-113706150 on Build GRCh38
rs199923434 Chr.1: 113706071-113706071 on Build GRCh38
rs867096370 Chr.1: 113706040-113706040 on Build GRCh38
rs761981459 Chr.1: 113706092-113706092 on Build GRCh38
rs761976536 Chr.1: 113706024-113706024 on Build GRCh38
rs753703552 Chr.1: 113706046-113706046 on Build GRCh38
rs756188876 Chr.1: 113706036-113706036 on Build GRCh38
rs778655762 Chr.1: 113706052-113706052 on Build GRCh38

View Full Product Details