Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 8039545-8039782 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ALOX15B
Entrez Gene ID
247
Gene Name
arachidonate 15-lipoxygenase, type B
Gene Aliases
15-LOX-2
UniGene
Hs.111256
Gene Chromosome Location
Chr.17: 8039040-8049134 on Build GRCh38
Transcripts
NM_001039130.1 NM_001039131.1 NM_001141.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6387713 c.359A>G p.E120G Chr.17: 8039597-8039597 on Build GRCh38
6538636 c.313C>G p.P105A Chr.17: 8039551-8039551 on Build GRCh38
5789229 c.367+8G>C p.? Chr.17: 8039613-8039613 on Build GRCh38
3749425 c.345C>G p.T115T Chr.17: 8039583-8039583 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs925454575 Chr.17: 8039664-8039664 on Build GRCh38
rs780267136 Chr.17: 8039613-8039613 on Build GRCh38
rs894772839 Chr.17: 8039586-8039586 on Build GRCh38
rs974104385 Chr.17: 8039758-8039758 on Build GRCh38
rs955234100 Chr.17: 8039669-8039669 on Build GRCh38
rs537711761 Chr.17: 8039657-8039657 on Build GRCh38
rs760405955 Chr.17: 8039568-8039568 on Build GRCh38
rs554229653 Chr.17: 8039760-8039760 on Build GRCh38
rs987099383 Chr.17: 8039672-8039672 on Build GRCh38
rs556160387 Chr.17: 8039660-8039660 on Build GRCh38
rs76589243 Chr.17: 8039583-8039583 on Build GRCh38
rs892724552 Chr.17: 8039762-8039762 on Build GRCh38
rs190700969 Chr.17: 8039723-8039723 on Build GRCh38
rs750640868 Chr.17: 8039590-8039590 on Build GRCh38
rs758759480 Chr.17: 8039610-8039610 on Build GRCh38
rs752493888 Chr.17: 8039567-8039567 on Build GRCh38
rs1018203958 Chr.17: 8039710-8039710 on Build GRCh38
rs755472977 Chr.17: 8039651-8039651 on Build GRCh38
rs751694404 Chr.17: 8039614-8039614 on Build GRCh38
rs571259142 Chr.17: 8039665-8039665 on Build GRCh38
rs182940148 Chr.17: 8039763-8039763 on Build GRCh38
rs1022422718 Chr.17: 8039611-8039611 on Build GRCh38
rs567205949 Chr.17: 8039646-8039646 on Build GRCh38

View Full Product Details