Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 9379045-9379280 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZNF177
Entrez Gene ID
7730
Gene Name
zinc finger protein 177
Gene Aliases
PIGX
UniGene
Hs.728944
Gene Chromosome Location
Chr.19: 9363020-9382617 on Build GRCh38
Transcripts
NM_001172651.1 NM_003451.2 NM_001172650.2 NM_001202425.1

Gene Symbol
ZNF559-ZNF177
Entrez Gene ID
100529215
Gene Name
ZNF559-ZNF177 readthrough
Gene Aliases
-
UniGene
Hs.728944
Gene Chromosome Location
Chr.19: 9324226-9382617 on Build GRCh38
Transcripts
NM_001172651.1 NM_003451.2 NM_001172650.2 NM_001202425.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3960844 c.121G>T p.D41Y Chr.19: 9379049-9379049 on Build GRCh38
242357 c.127A>C p.M43L Chr.19: 9379055-9379055 on Build GRCh38
242358 c.127A>C p.M43L Chr.19: 9379055-9379055 on Build GRCh38
7320596 c.126G>A p.V42V Chr.19: 9379054-9379054 on Build GRCh38
3960843 c.121G>T p.D41Y Chr.19: 9379049-9379049 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1056417632 Chr.19: 9379153-9379153 on Build GRCh38
rs138231601 Chr.19: 9379166-9379166 on Build GRCh38
rs185343874 Chr.19: 9379160-9379160 on Build GRCh38
rs930335315 Chr.19: 9379177-9379177 on Build GRCh38
rs778104819 Chr.19: 9379104-9379104 on Build GRCh38
rs770547727 Chr.19: 9379140-9379140 on Build GRCh38
rs1003783838 Chr.19: 9379113-9379113 on Build GRCh38
rs748977959 Chr.19: 9379135-9379135 on Build GRCh38
rs912081091 Chr.19: 9379148-9379148 on Build GRCh38
rs920220046 Chr.19: 9379174-9379174 on Build GRCh38
rs886297088 Chr.19: 9379103-9379103 on Build GRCh38
rs537575840 Chr.19: 9379202-9379202 on Build GRCh38
rs939150077 Chr.19: 9379221-9379221 on Build GRCh38
rs1044831677 Chr.19: 9379186-9379186 on Build GRCh38
rs756490669 Chr.19: 9379091-9379091 on Build GRCh38
rs775601195 Chr.19: 9379075-9379075 on Build GRCh38
rs3745575 Chr.19: 9379210-9379210 on Build GRCh38

View Full Product Details