Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 1323690-1323941 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CLPTM1L
Entrez Gene ID
81037
Gene Name
CLPTM1 like
Gene Aliases
CRR9
UniGene
Hs.444673
Gene Chromosome Location
Chr.5: 1317744-1345070 on Build GRCh38
Transcripts
NM_030782.4 XM_011514144.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5912201 c.1198-7C>T p.? Chr.5: 1323876-1323876 on Build GRCh38
7241322 c.1274A>G p.Y425C Chr.5: 1323793-1323793 on Build GRCh38
735688 c.1276A>T p.K426* Chr.5: 1323791-1323791 on Build GRCh38
5709455 c.1240G>A p.G414R Chr.5: 1323827-1323827 on Build GRCh38
4462270 c.1239C>T p.V413V Chr.5: 1323828-1323828 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs202162399 Chr.5: 1323805-1323805 on Build GRCh38
rs745497515 Chr.5: 1323777-1323777 on Build GRCh38
rs375610957 Chr.5: 1323807-1323807 on Build GRCh38
rs369488069 Chr.5: 1323772-1323772 on Build GRCh38
rs766018144 Chr.5: 1323823-1323823 on Build GRCh38
rs747095800 Chr.5: 1323875-1323875 on Build GRCh38
rs749656094 Chr.5: 1323760-1323760 on Build GRCh38
rs775621154 Chr.5: 1323865-1323865 on Build GRCh38
rs1056403986 Chr.5: 1323818-1323818 on Build GRCh38
rs140752465 Chr.5: 1323726-1323726 on Build GRCh38
rs773244135 Chr.5: 1323916-1323916 on Build GRCh38
rs200291959 Chr.5: 1323830-1323830 on Build GRCh38
rs747859153 Chr.5: 1323917-1323917 on Build GRCh38
rs559687363 Chr.5: 1323715-1323715 on Build GRCh38
rs780776403 Chr.5: 1323832-1323832 on Build GRCh38
rs373545372 Chr.5: 1323776-1323776 on Build GRCh38
rs576070650 Chr.5: 1323718-1323718 on Build GRCh38
rs369954976 Chr.5: 1323825-1323825 on Build GRCh38
rs772092980 Chr.5: 1323857-1323857 on Build GRCh38
rs779329573 Chr.5: 1323843-1323843 on Build GRCh38
rs778395654 Chr.5: 1323902-1323902 on Build GRCh38
rs117731060 Chr.5: 1323756-1323756 on Build GRCh38
rs139068003 Chr.5: 1323814-1323814 on Build GRCh38
rs376967202 Chr.5: 1323909-1323909 on Build GRCh38
rs764275631 Chr.5: 1323793-1323793 on Build GRCh38
rs761640182 Chr.5: 1323903-1323903 on Build GRCh38
rs201262528 Chr.5: 1323757-1323757 on Build GRCh38
rs199811270 Chr.5: 1323771-1323771 on Build GRCh38
rs143020533 Chr.5: 1323821-1323821 on Build GRCh38
rs3817067 Chr.5: 1323731-1323731 on Build GRCh38
rs768916470 Chr.5: 1323892-1323892 on Build GRCh38
rs772148054 Chr.5: 1323774-1323774 on Build GRCh38
rs760981483 Chr.5: 1323780-1323780 on Build GRCh38
rs765249449 Chr.5: 1323810-1323810 on Build GRCh38
rs776771733 Chr.5: 1323899-1323899 on Build GRCh38
rs985173029 Chr.5: 1323734-1323734 on Build GRCh38
rs200310656 Chr.5: 1323769-1323769 on Build GRCh38
rs749652158 Chr.5: 1323840-1323840 on Build GRCh38
rs554612323 Chr.5: 1323708-1323708 on Build GRCh38
rs746408773 Chr.5: 1323851-1323851 on Build GRCh38
rs375045987 Chr.5: 1323770-1323770 on Build GRCh38

View Full Product Details