Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 74038407-74038650 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CXCL3
Entrez Gene ID
2921
Gene Name
C-X-C motif chemokine ligand 3
Gene Aliases
CINC-2b, GRO3, GROg, MIP-2b, MIP2B, SCYB3
UniGene
Hs.89690
Gene Chromosome Location
Chr.4: 74036589-74038773 on Build GRCh38
Transcripts
NM_002090.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3965572 c.2T>C p.M1T Chr.4: 74038610-74038610 on Build GRCh38
5890009 c.100+7C>T p.? Chr.4: 74038505-74038505 on Build GRCh38
4159170 c.21C>A p.S7S Chr.4: 74038591-74038591 on Build GRCh38
3760733 c.33C>A p.S11R Chr.4: 74038579-74038579 on Build GRCh38
1057483 c.105G>A p.A35A Chr.4: 74038409-74038409 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs749012465 Chr.4: 74038503-74038503 on Build GRCh38
rs559863732 Chr.4: 74038561-74038561 on Build GRCh38
rs352043 Chr.4: 74038604-74038604 on Build GRCh38
rs199994419 Chr.4: 74038451-74038451 on Build GRCh38
rs770596414 Chr.4: 74038432-74038432 on Build GRCh38
rs1040300007 Chr.4: 74038447-74038447 on Build GRCh38
rs371114059 Chr.4: 74038454-74038454 on Build GRCh38
rs530265218 Chr.4: 74038452-74038452 on Build GRCh38
rs1030785182 Chr.4: 74038538-74038538 on Build GRCh38
rs189466801 Chr.4: 74038563-74038563 on Build GRCh38
rs960638906 Chr.4: 74038540-74038540 on Build GRCh38
rs375665200 Chr.4: 74038471-74038471 on Build GRCh38
rs879217871 Chr.4: 74038510-74038510 on Build GRCh38
rs773370929 Chr.4: 74038505-74038505 on Build GRCh38
rs550117892 Chr.4: 74038607-74038607 on Build GRCh38
rs751227846 Chr.4: 74038567-74038567 on Build GRCh38
rs780606476 Chr.4: 74038477-74038477 on Build GRCh38
rs753759622 Chr.4: 74038434-74038437 on Build GRCh38
rs768467893 Chr.4: 74038504-74038504 on Build GRCh38
rs570431901 Chr.4: 74038601-74038601 on Build GRCh38
rs200514475 Chr.4: 74038431-74038431 on Build GRCh38
rs756807505 Chr.4: 74038491-74038491 on Build GRCh38
rs951616904 Chr.4: 74038596-74038596 on Build GRCh38
rs765625566 Chr.4: 74038468-74038468 on Build GRCh38
rs1022150215 Chr.4: 74038519-74038519 on Build GRCh38
rs182216932 Chr.4: 74038579-74038579 on Build GRCh38
rs769677798 Chr.4: 74038485-74038485 on Build GRCh38
rs754341806 Chr.4: 74038464-74038464 on Build GRCh38
rs765601099 Chr.4: 74038559-74038559 on Build GRCh38
rs777528839 Chr.4: 74038474-74038474 on Build GRCh38
rs756808288 Chr.4: 74038475-74038475 on Build GRCh38
rs202236627 Chr.4: 74038480-74038480 on Build GRCh38
rs901358285 Chr.4: 74038448-74038448 on Build GRCh38
rs775510991 Chr.4: 74038500-74038500 on Build GRCh38
rs201483717 Chr.4: 74038437-74038437 on Build GRCh38
rs769599022 Chr.4: 74038439-74038439 on Build GRCh38
rs903566310 Chr.4: 74038522-74038522 on Build GRCh38
rs760933758 Chr.4: 74038508-74038508 on Build GRCh38
rs776634963 Chr.4: 74038521-74038521 on Build GRCh38
rs199829667 Chr.4: 74038555-74038555 on Build GRCh38
rs568243232 Chr.4: 74038625-74038625 on Build GRCh38
rs935953679 Chr.4: 74038611-74038611 on Build GRCh38
rs745537791 Chr.4: 74038450-74038451 on Build GRCh38
rs999680796 Chr.4: 74038529-74038529 on Build GRCh38
rs201088403 Chr.4: 74038466-74038466 on Build GRCh38
rs995986907 Chr.4: 74038492-74038492 on Build GRCh38
rs910238744 Chr.4: 74038628-74038628 on Build GRCh38
rs766388869 Chr.4: 74038462-74038462 on Build GRCh38
rs1034387222 Chr.4: 74038482-74038482 on Build GRCh38
rs766477754 Chr.4: 74038514-74038514 on Build GRCh38
rs974180316 Chr.4: 74038572-74038572 on Build GRCh38
rs200464221 Chr.4: 74038602-74038602 on Build GRCh38
rs958682864 Chr.4: 74038489-74038489 on Build GRCh38
rs760702836 Chr.4: 74038457-74038457 on Build GRCh38

View Full Product Details