Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 21092147-21092396 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZNF219
Entrez Gene ID
51222
Gene Name
zinc finger protein 219
Gene Aliases
ZFP219
UniGene
Hs.250493
Gene Chromosome Location
Chr.14: 21090046-21104704 on Build GRCh38
Transcripts
NM_001101672.1 NM_001102454.1 NM_016423.2 XM_005267739.1 XM_006720163.2 XM_006720164.3 XM_011536825.1 XM_017021354.1 XM_017021355.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3690010 c.936G>T p.K312N Chr.14: 21092361-21092361 on Build GRCh38
4600358 c.922G>A p.G308S Chr.14: 21092375-21092375 on Build GRCh38
4147976 c.1050C>T p.G350G Chr.14: 21092247-21092247 on Build GRCh38
5964994 c.1111G>A p.E371K Chr.14: 21092186-21092186 on Build GRCh38
4890526 c.994C>A p.R332S Chr.14: 21092303-21092303 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs769554730 Chr.14: 21092249-21092249 on Build GRCh38
rs371511967 Chr.14: 21092271-21092271 on Build GRCh38
rs896186659 Chr.14: 21092262-21092262 on Build GRCh38
rs752651846 Chr.14: 21092375-21092375 on Build GRCh38
rs921348586 Chr.14: 21092191-21092191 on Build GRCh38
rs991441624 Chr.14: 21092166-21092166 on Build GRCh38
rs1048448941 Chr.14: 21092203-21092203 on Build GRCh38
rs774310193 Chr.14: 21092277-21092277 on Build GRCh38
rs998642276 Chr.14: 21092367-21092367 on Build GRCh38
rs772773620 Chr.14: 21092287-21092287 on Build GRCh38
rs904962693 Chr.14: 21092276-21092276 on Build GRCh38
rs888765056 Chr.14: 21092274-21092274 on Build GRCh38
rs199878395 Chr.14: 21092296-21092296 on Build GRCh38
rs766037506 Chr.14: 21092320-21092320 on Build GRCh38
rs868270275 Chr.14: 21092371-21092371 on Build GRCh38
rs902953539 Chr.14: 21092316-21092316 on Build GRCh38
rs908631359 Chr.14: 21092211-21092211 on Build GRCh38
rs747976313 Chr.14: 21092247-21092247 on Build GRCh38
rs767490416 Chr.14: 21092373-21092373 on Build GRCh38
rs931371939 Chr.14: 21092202-21092202 on Build GRCh38
rs1013265382 Chr.14: 21092269-21092269 on Build GRCh38
rs776113024 Chr.14: 21092290-21092290 on Build GRCh38
rs751094498 Chr.14: 21092322-21092322 on Build GRCh38
rs887593110 Chr.14: 21092246-21092246 on Build GRCh38
rs762522847 Chr.14: 21092305-21092305 on Build GRCh38
rs35548569 Chr.14: 21092272-21092272 on Build GRCh38
rs1021362624 Chr.14: 21092307-21092307 on Build GRCh38
rs957075642 Chr.14: 21092324-21092324 on Build GRCh38
rs758803894 Chr.14: 21092283-21092283 on Build GRCh38
rs777573435 Chr.14: 21092250-21092250 on Build GRCh38
rs761172534 Chr.14: 21092293-21092293 on Build GRCh38
rs867786813 Chr.14: 21092342-21092342 on Build GRCh38
rs777018373 Chr.14: 21092302-21092302 on Build GRCh38
rs138145521 Chr.14: 21092270-21092270 on Build GRCh38
rs866157997 Chr.14: 21092341-21092341 on Build GRCh38
rs372627398 Chr.14: 21092231-21092231 on Build GRCh38
rs1048892886 Chr.14: 21092229-21092229 on Build GRCh38
rs754449837 Chr.14: 21092358-21092358 on Build GRCh38
rs554406948 Chr.14: 21092179-21092179 on Build GRCh38

View Full Product Details