Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 58491582-58491815 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CYP7A1
Entrez Gene ID
1581
Gene Name
cytochrome P450 family 7 subfamily A member 1
Gene Aliases
CP7A, CYP7, CYPVII
UniGene
Hs.1644
Gene Chromosome Location
Chr.8: 58490178-58500161 on Build GRCh38
Transcripts
NM_000780.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3735320 c.1319G>A p.G440E Chr.8: 58491671-58491671 on Build GRCh38
2719710 c.1357G>A p.E453K Chr.8: 58491633-58491633 on Build GRCh38
7363579 c.1316C>T p.S439L Chr.8: 58491674-58491674 on Build GRCh38
6639493 c.1328T>C p.I443T Chr.8: 58491662-58491662 on Build GRCh38
486552 c.1239T>G p.L413L Chr.8: 58491751-58491751 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs776760317 Chr.8: 58491712-58491712 on Build GRCh38
rs755342899 Chr.8: 58491633-58491633 on Build GRCh38
rs762092457 Chr.8: 58491649-58491649 on Build GRCh38
rs768925733 Chr.8: 58491708-58491708 on Build GRCh38
rs777507940 Chr.8: 58491754-58491754 on Build GRCh38
rs868642976 Chr.8: 58491775-58491775 on Build GRCh38
rs192086535 Chr.8: 58491625-58491625 on Build GRCh38
rs765527179 Chr.8: 58491718-58491718 on Build GRCh38
rs772113117 Chr.8: 58491778-58491778 on Build GRCh38
rs748282246 Chr.8: 58491781-58491781 on Build GRCh38
rs770804582 Chr.8: 58491690-58491690 on Build GRCh38
rs780834574 Chr.8: 58491612-58491612 on Build GRCh38
rs760617117 Chr.8: 58491739-58491739 on Build GRCh38
rs367898327 Chr.8: 58491671-58491671 on Build GRCh38
rs976831279 Chr.8: 58491639-58491639 on Build GRCh38
rs754119848 Chr.8: 58491647-58491647 on Build GRCh38
rs769540638 Chr.8: 58491632-58491632 on Build GRCh38
rs776614332 Chr.8: 58491780-58491780 on Build GRCh38
rs8192878 Chr.8: 58491787-58491787 on Build GRCh38
rs568116014 Chr.8: 58491736-58491736 on Build GRCh38
rs201341987 Chr.8: 58491686-58491686 on Build GRCh38
rs899918229 Chr.8: 58491688-58491688 on Build GRCh38
rs750778526 Chr.8: 58491656-58491656 on Build GRCh38
rs201787113 Chr.8: 58491741-58491741 on Build GRCh38
rs964395133 Chr.8: 58491634-58491634 on Build GRCh38
rs778848469 Chr.8: 58491766-58491766 on Build GRCh38
rs773486919 Chr.8: 58491789-58491789 on Build GRCh38
rs891054570 Chr.8: 58491614-58491614 on Build GRCh38
rs752679173 Chr.8: 58491715-58491715 on Build GRCh38
rs773066134 Chr.8: 58491738-58491739 on Build GRCh38
rs546561123 Chr.8: 58491744-58491744 on Build GRCh38
rs764215156 Chr.8: 58491646-58491646 on Build GRCh38
rs780673203 Chr.8: 58491674-58491674 on Build GRCh38
rs747810583 Chr.8: 58491681-58491681 on Build GRCh38
rs745860537 Chr.8: 58491773-58491773 on Build GRCh38
rs766284289 Chr.8: 58491751-58491752 on Build GRCh38
rs552280210 Chr.8: 58491663-58491663 on Build GRCh38
rs915543380 Chr.8: 58491724-58491724 on Build GRCh38
rs747766938 Chr.8: 58491616-58491616 on Build GRCh38
rs752124663 Chr.8: 58491734-58491734 on Build GRCh38
rs757124756 Chr.8: 58491762-58491762 on Build GRCh38
rs773258821 Chr.8: 58491725-58491725 on Build GRCh38
rs114370904 Chr.8: 58491742-58491742 on Build GRCh38
rs1027036041 Chr.8: 58491719-58491719 on Build GRCh38
rs770873945 Chr.8: 58491642-58491642 on Build GRCh38
rs532694162 Chr.8: 58491733-58491733 on Build GRCh38
rs748837433 Chr.8: 58491757-58491757 on Build GRCh38
rs878898857 Chr.8: 58491666-58491666 on Build GRCh38
rs370236722 Chr.8: 58491697-58491697 on Build GRCh38
rs554746594 Chr.8: 58491730-58491730 on Build GRCh38
rs1030176083 Chr.8: 58491664-58491664 on Build GRCh38
rs777464708 Chr.8: 58491689-58491689 on Build GRCh38
rs151122002 Chr.8: 58491698-58491698 on Build GRCh38
rs376754937 Chr.8: 58491705-58491705 on Build GRCh38
rs771884497 Chr.8: 58491699-58491703 on Build GRCh38
rs925861393 Chr.8: 58491691-58491691 on Build GRCh38
rs754486556 Chr.8: 58491673-58491673 on Build GRCh38
rs199557345 Chr.8: 58491641-58491641 on Build GRCh38

View Full Product Details