Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 132748430-132748655 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCNI2
Entrez Gene ID
645121
Gene Name
cyclin I family member 2
Gene Aliases
-
UniGene
Hs.522057
Gene Chromosome Location
Chr.5: 132747402-132754403 on Build GRCh38
Transcripts
NM_001039780.3 NM_001287252.1 NM_001287253.1 XM_017009725.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
220231 c.526A>G p.T176A Chr.5: 132748443-132748443 on Build GRCh38
1753961 c.521C>T p.A174V Chr.5: 132748438-132748438 on Build GRCh38
5642595 c.517C>G p.L173V Chr.5: 132748434-132748434 on Build GRCh38
4796116 c.520G>A p.A174T Chr.5: 132748437-132748437 on Build GRCh38
243542 c.555G>T p.V185V Chr.5: 132748472-132748472 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs754823405 Chr.5: 132748497-132748497 on Build GRCh38
rs748208070 Chr.5: 132748485-132748485 on Build GRCh38
rs972213983 Chr.5: 132748533-132748533 on Build GRCh38
rs748056376 Chr.5: 132748517-132748517 on Build GRCh38
rs201156649 Chr.5: 132748456-132748456 on Build GRCh38
rs1056596248 Chr.5: 132748602-132748602 on Build GRCh38
rs1016401868 Chr.5: 132748480-132748480 on Build GRCh38
rs780401710 Chr.5: 132748455-132748455 on Build GRCh38
rs981775877 Chr.5: 132748578-132748578 on Build GRCh38
rs762853529 Chr.5: 132748498-132748498 on Build GRCh38
rs778710687 Chr.5: 132748502-132748502 on Build GRCh38
rs930626898 Chr.5: 132748536-132748536 on Build GRCh38
rs939469679 Chr.5: 132748601-132748601 on Build GRCh38
rs907956435 Chr.5: 132748600-132748600 on Build GRCh38
rs920572502 Chr.5: 132748534-132748534 on Build GRCh38
rs72799426 Chr.5: 132748537-132748537 on Build GRCh38
rs970779903 Chr.5: 132748566-132748566 on Build GRCh38

View Full Product Details