Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 114236545-114236798 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZNF80
Entrez Gene ID
7634
Gene Name
zinc finger protein 80
Gene Aliases
pT17
UniGene
Hs.271079
Gene Chromosome Location
Chr.3: 114211035-114237578 on Build GRCh38
Transcripts
NM_007136.3 XM_017007132.1 XM_017007133.1 XM_017007134.1 XM_017007135.1 XM_017007136.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6880074 c.339C>T p.V113V Chr.3: 114236736-114236736 on Build GRCh38
1484466 c.407G>A p.S136N Chr.3: 114236668-114236668 on Build GRCh38
372246 c.463C>A p.H155N Chr.3: 114236612-114236612 on Build GRCh38
3769725 c.525C>T p.S175S Chr.3: 114236550-114236550 on Build GRCh38
316914 c.451C>A p.H151N Chr.3: 114236624-114236624 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs928392764 Chr.3: 114236654-114236654 on Build GRCh38
rs968161742 Chr.3: 114236594-114236594 on Build GRCh38
rs1056037096 Chr.3: 114236681-114236681 on Build GRCh38
rs868491475 Chr.3: 114236621-114236621 on Build GRCh38
rs545841246 Chr.3: 114236753-114236753 on Build GRCh38
rs756461881 Chr.3: 114236613-114236613 on Build GRCh38
rs775794096 Chr.3: 114236688-114236688 on Build GRCh38
rs866579682 Chr.3: 114236675-114236675 on Build GRCh38
rs779304062 Chr.3: 114236755-114236755 on Build GRCh38
rs916528251 Chr.3: 114236645-114236645 on Build GRCh38
rs201704376 Chr.3: 114236685-114236685 on Build GRCh38
rs766667293 Chr.3: 114236720-114236720 on Build GRCh38
rs759155465 Chr.3: 114236722-114236722 on Build GRCh38
rs753494488 Chr.3: 114236624-114236624 on Build GRCh38
rs761034353 Chr.3: 114236719-114236719 on Build GRCh38
rs777690622 Chr.3: 114236754-114236755 on Build GRCh38
rs774544663 Chr.3: 114236704-114236704 on Build GRCh38
rs200366369 Chr.3: 114236728-114236728 on Build GRCh38
rs769773258 Chr.3: 114236701-114236702 on Build GRCh38
rs1023476800 Chr.3: 114236771-114236771 on Build GRCh38
rs572397764 Chr.3: 114236747-114236747 on Build GRCh38
rs777695922 Chr.3: 114236730-114236730 on Build GRCh38
rs140075528 Chr.3: 114236766-114236766 on Build GRCh38
rs147292609 Chr.3: 114236612-114236612 on Build GRCh38
rs139382869 Chr.3: 114236667-114236667 on Build GRCh38
rs774628469 Chr.3: 114236592-114236592 on Build GRCh38
rs770632220 Chr.3: 114236687-114236687 on Build GRCh38
rs541896176 Chr.3: 114236680-114236680 on Build GRCh38
rs746774079 Chr.3: 114236686-114236686 on Build GRCh38
rs375580832 Chr.3: 114236590-114236590 on Build GRCh38
rs749505351 Chr.3: 114236690-114236690 on Build GRCh38
rs773623409 Chr.3: 114236716-114236716 on Build GRCh38
rs560258157 Chr.3: 114236732-114236732 on Build GRCh38
rs778382959 Chr.3: 114236656-114236656 on Build GRCh38
rs148508596 Chr.3: 114236744-114236744 on Build GRCh38
rs142751789 Chr.3: 114236752-114236752 on Build GRCh38
rs767952335 Chr.3: 114236596-114236596 on Build GRCh38
rs748223184 Chr.3: 114236760-114236760 on Build GRCh38
rs140260405 Chr.3: 114236642-114236642 on Build GRCh38
rs768023432 Chr.3: 114236710-114236710 on Build GRCh38
rs753305973 Chr.3: 114236770-114236770 on Build GRCh38
rs555903765 Chr.3: 114236579-114236579 on Build GRCh38
rs143414946 Chr.3: 114236776-114236776 on Build GRCh38
rs903552852 Chr.3: 114236749-114236749 on Build GRCh38
rs766015106 Chr.3: 114236620-114236620 on Build GRCh38
rs370641478 Chr.3: 114236705-114236705 on Build GRCh38
rs769068910 Chr.3: 114236692-114236692 on Build GRCh38
rs960569132 Chr.3: 114236639-114236639 on Build GRCh38
rs919947624 Chr.3: 114236691-114236691 on Build GRCh38
rs757877383 Chr.3: 114236729-114236729 on Build GRCh38
rs762407180 Chr.3: 114236593-114236593 on Build GRCh38
rs772050375 Chr.3: 114236761-114236761 on Build GRCh38
rs1030779342 Chr.3: 114236757-114236757 on Build GRCh38
rs145218257 Chr.3: 114236765-114236765 on Build GRCh38
rs144823946 Chr.3: 114236721-114236721 on Build GRCh38
rs143330045 Chr.3: 114236677-114236677 on Build GRCh38
rs764278880 Chr.3: 114236582-114236582 on Build GRCh38
rs1011832459 Chr.3: 114236745-114236745 on Build GRCh38

View Full Product Details