Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 72905849-72906118 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ALDH1A1
Entrez Gene ID
216
Gene Name
aldehyde dehydrogenase 1 family member A1
Gene Aliases
ALDC, ALDH-E1, ALDH1, ALDH11, HEL-9, HEL-S-53e, HEL12, PUMB1, RALDH1
UniGene
Hs.76392
Gene Chromosome Location
Chr.9: 72900662-72953317 on Build GRCh38
Transcripts
NM_000689.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5411230 c.1362G>A p.V454V Chr.9: 72906029-72906029 on Build GRCh38
5900528 c.1359-7C>T p.? Chr.9: 72906039-72906039 on Build GRCh38
3930103 c.1373G>A p.G458D Chr.9: 72906018-72906018 on Build GRCh38
1283371 c.1373G>C p.G458A Chr.9: 72906018-72906018 on Build GRCh38
4465686 c.1395C>T p.P465P Chr.9: 72905996-72905996 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs3739960 Chr.9: 72905941-72905941 on Build GRCh38
rs764380819 Chr.9: 72905948-72905948 on Build GRCh38
rs774366233 Chr.9: 72905930-72905930 on Build GRCh38
rs763363983 Chr.9: 72906013-72906013 on Build GRCh38
rs8187985 Chr.9: 72906045-72906049 on Build GRCh38
rs770639291 Chr.9: 72906050-72906051 on Build GRCh38
rs1063447 Chr.9: 72905994-72905994 on Build GRCh38
rs771973267 Chr.9: 72905946-72905946 on Build GRCh38
rs908840133 Chr.9: 72905906-72905906 on Build GRCh38
rs879501142 Chr.9: 72906059-72906059 on Build GRCh38
rs201658437 Chr.9: 72905951-72905951 on Build GRCh38
rs776954405 Chr.9: 72905949-72905949 on Build GRCh38
rs773576868 Chr.9: 72905918-72905923 on Build GRCh38
rs766918121 Chr.9: 72905912-72905912 on Build GRCh38
rs1008773313 Chr.9: 72905999-72905999 on Build GRCh38
rs765238490 Chr.9: 72906050-72906050 on Build GRCh38
rs370163540 Chr.9: 72905924-72905924 on Build GRCh38
rs997456207 Chr.9: 72905943-72905943 on Build GRCh38
rs530247413 Chr.9: 72906077-72906077 on Build GRCh38
rs755799729 Chr.9: 72906017-72906017 on Build GRCh38
rs750942320 Chr.9: 72905978-72905978 on Build GRCh38
rs944301739 Chr.9: 72905917-72905917 on Build GRCh38
rs765175005 Chr.9: 72905969-72905969 on Build GRCh38
rs756770659 Chr.9: 72906053-72906053 on Build GRCh38
rs753437277 Chr.9: 72906045-72906045 on Build GRCh38
rs771885118 Chr.9: 72906084-72906084 on Build GRCh38
rs755750614 Chr.9: 72905913-72905913 on Build GRCh38
rs770385981 Chr.9: 72905928-72905928 on Build GRCh38
rs143437440 Chr.9: 72906016-72906016 on Build GRCh38
rs1053219143 Chr.9: 72905889-72905889 on Build GRCh38
rs540688031 Chr.9: 72906039-72906043 on Build GRCh38
rs776417057 Chr.9: 72906076-72906077 on Build GRCh38
rs778285139 Chr.9: 72906062-72906062 on Build GRCh38
rs777430527 Chr.9: 72906029-72906029 on Build GRCh38
rs959908011 Chr.9: 72906057-72906057 on Build GRCh38
rs758998877 Chr.9: 72906031-72906031 on Build GRCh38
rs200569776 Chr.9: 72905947-72905947 on Build GRCh38
rs745878761 Chr.9: 72906077-72906077 on Build GRCh38

View Full Product Details