Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 43515107-43515350 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
L3MBTL1
Entrez Gene ID
26013
Gene Name
l(3)mbt-like 1 (Drosophila)
Gene Aliases
H-L(3)MBT, L3MBTL, ZC2HC3, dJ138B7.3
UniGene
Hs.709356
Gene Chromosome Location
Chr.20: 43507680-43541895 on Build GRCh38
Transcripts
NM_015478.6 NM_032107.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4697143 c.340C>A p.L114I Chr.20: 43515116-43515116 on Build GRCh38
6354684 c.631G>T p.E211* Chr.20: 43515335-43515335 on Build GRCh38
4499294 c.539C>T p.P180L Chr.20: 43515111-43515111 on Build GRCh38
6565844 c.367C>G p.Q123E Chr.20: 43515143-43515143 on Build GRCh38
7130008 c.579C>G p.F193L Chr.20: 43515151-43515151 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs760876091 Chr.20: 43515181-43515181 on Build GRCh38
rs759637086 Chr.20: 43515159-43515159 on Build GRCh38
rs777752080 Chr.20: 43515140-43515140 on Build GRCh38
rs926593400 Chr.20: 43515221-43515221 on Build GRCh38
rs185699630 Chr.20: 43515283-43515283 on Build GRCh38
rs747703276 Chr.20: 43515135-43515135 on Build GRCh38
rs767199298 Chr.20: 43515211-43515211 on Build GRCh38
rs181047062 Chr.20: 43515240-43515240 on Build GRCh38
rs957051045 Chr.20: 43515291-43515291 on Build GRCh38
rs754524487 Chr.20: 43515196-43515196 on Build GRCh38
rs993181243 Chr.20: 43515171-43515171 on Build GRCh38
rs768899273 Chr.20: 43515299-43515299 on Build GRCh38
rs375417824 Chr.20: 43515210-43515210 on Build GRCh38
rs368664102 Chr.20: 43515137-43515137 on Build GRCh38
rs749262874 Chr.20: 43515257-43515257 on Build GRCh38
rs866848965 Chr.20: 43515217-43515217 on Build GRCh38
rs752141763 Chr.20: 43515241-43515241 on Build GRCh38
rs950235582 Chr.20: 43515315-43515315 on Build GRCh38
rs201768979 Chr.20: 43515255-43515255 on Build GRCh38
rs765660061 Chr.20: 43515190-43515190 on Build GRCh38
rs777594692 Chr.20: 43515131-43515131 on Build GRCh38
rs145351923 Chr.20: 43515271-43515271 on Build GRCh38
rs560016568 Chr.20: 43515256-43515256 on Build GRCh38
rs149664498 Chr.20: 43515144-43515144 on Build GRCh38
rs765856381 Chr.20: 43515313-43515313 on Build GRCh38
rs200368697 Chr.20: 43515164-43515164 on Build GRCh38
rs775840279 Chr.20: 43515295-43515295 on Build GRCh38
rs770545404 Chr.20: 43515189-43515190 on Build GRCh38
rs774524586 Chr.20: 43515158-43515158 on Build GRCh38
rs762364128 Chr.20: 43515308-43515308 on Build GRCh38
rs775366251 Chr.20: 43515166-43515166 on Build GRCh38
rs940191570 Chr.20: 43515289-43515289 on Build GRCh38
rs199582590 Chr.20: 43515254-43515254 on Build GRCh38
rs757216190 Chr.20: 43515265-43515265 on Build GRCh38
rs201779402 Chr.20: 43515182-43515182 on Build GRCh38
rs533065169 Chr.20: 43515226-43515226 on Build GRCh38
rs368713154 Chr.20: 43515209-43515209 on Build GRCh38
rs750825655 Chr.20: 43515192-43515192 on Build GRCh38
rs763438994 Chr.20: 43515328-43515328 on Build GRCh38
rs757200960 Chr.20: 43515180-43515180 on Build GRCh38
rs762239461 Chr.20: 43515184-43515184 on Build GRCh38
rs531085763 Chr.20: 43515132-43515132 on Build GRCh38
rs767060691 Chr.20: 43515329-43515329 on Build GRCh38
rs200165694 Chr.20: 43515188-43515188 on Build GRCh38
rs999940470 Chr.20: 43515141-43515141 on Build GRCh38
rs773378177 Chr.20: 43515325-43515325 on Build GRCh38
rs745680399 Chr.20: 43515275-43515275 on Build GRCh38
rs754447208 Chr.20: 43515183-43515183 on Build GRCh38
rs908729424 Chr.20: 43515284-43515284 on Build GRCh38
rs770760224 Chr.20: 43515157-43515157 on Build GRCh38
rs964960199 Chr.20: 43515239-43515239 on Build GRCh38
rs145435727 Chr.20: 43515296-43515296 on Build GRCh38
rs555402988 Chr.20: 43515282-43515282 on Build GRCh38
rs8183462 Chr.20: 43515320-43515320 on Build GRCh38
rs776885081 Chr.20: 43515302-43515302 on Build GRCh38

View Full Product Details