Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 35860879-35861104 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KIRREL2
Entrez Gene ID
84063
Gene Name
kin of IRRE like 2 (Drosophila)
Gene Aliases
FILTRIN, NEPH3, NLG1
UniGene
Hs.145729
Gene Chromosome Location
Chr.19: 35851635-35867146 on Build GRCh38
Transcripts
NM_032123.6 NM_199179.3 NM_199180.3 XM_011527362.1 XM_011527363.1 XM_011527365.1 XM_011527366.1 XM_017027348.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
123990 c.1007G>A p.R336H Chr.19: 35860987-35860987 on Build GRCh38
4881423 c.933G>T p.G311G Chr.19: 35860913-35860913 on Build GRCh38
3404147 c.961G>A p.V321M Chr.19: 35860941-35860941 on Build GRCh38
4562640 c.936G>A p.P312P Chr.19: 35860916-35860916 on Build GRCh38
5367972 c.974_975insG p.E327fs*52 Chr.19: 35860954-35860955 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs775542301 Chr.19: 35860997-35860997 on Build GRCh38
rs757276599 Chr.19: 35860939-35860939 on Build GRCh38
rs774209299 Chr.19: 35861006-35861006 on Build GRCh38
rs773969029 Chr.19: 35861074-35861074 on Build GRCh38
rs755656040 Chr.19: 35861022-35861022 on Build GRCh38
rs768926805 Chr.19: 35861072-35861072 on Build GRCh38
rs746410812 Chr.19: 35861042-35861042 on Build GRCh38
rs769907828 Chr.19: 35860989-35860989 on Build GRCh38
rs553235458 Chr.19: 35861013-35861013 on Build GRCh38
rs376014818 Chr.19: 35861011-35861011 on Build GRCh38
rs776648314 Chr.19: 35860982-35860982 on Build GRCh38
rs377661393 Chr.19: 35861023-35861023 on Build GRCh38
rs758688884 Chr.19: 35860965-35860965 on Build GRCh38
rs762630821 Chr.19: 35860922-35860922 on Build GRCh38
rs201741181 Chr.19: 35860969-35860969 on Build GRCh38
rs371315188 Chr.19: 35861037-35861037 on Build GRCh38
rs774439246 Chr.19: 35860911-35860911 on Build GRCh38
rs554279522 Chr.19: 35861040-35861040 on Build GRCh38
rs377220880 Chr.19: 35860964-35860964 on Build GRCh38
rs370488330 Chr.19: 35860980-35860980 on Build GRCh38
rs891646141 Chr.19: 35860983-35860983 on Build GRCh38
rs767462367 Chr.19: 35861078-35861078 on Build GRCh38
rs753421630 Chr.19: 35861026-35861026 on Build GRCh38
rs889422633 Chr.19: 35860938-35860938 on Build GRCh38
rs73928339 Chr.19: 35861058-35861058 on Build GRCh38
rs763988199 Chr.19: 35860925-35860925 on Build GRCh38
rs373178887 Chr.19: 35860993-35860993 on Build GRCh38
rs745966676 Chr.19: 35860986-35860986 on Build GRCh38
rs779832976 Chr.19: 35860960-35860960 on Build GRCh38
rs761617260 Chr.19: 35861009-35861009 on Build GRCh38
rs905602190 Chr.19: 35861053-35861053 on Build GRCh38
rs768135613 Chr.19: 35861073-35861073 on Build GRCh38
rs749984753 Chr.19: 35860951-35860951 on Build GRCh38
rs543176004 Chr.19: 35861069-35861069 on Build GRCh38
rs200879020 Chr.19: 35860998-35860998 on Build GRCh38
rs752437508 Chr.19: 35860921-35860921 on Build GRCh38
rs1034099760 Chr.19: 35861068-35861068 on Build GRCh38
rs35854130 Chr.19: 35861033-35861033 on Build GRCh38
rs751445902 Chr.19: 35860932-35860932 on Build GRCh38
rs187468812 Chr.19: 35860959-35860959 on Build GRCh38
rs763300598 Chr.19: 35861002-35861002 on Build GRCh38
rs762169563 Chr.19: 35860994-35860994 on Build GRCh38
rs767077893 Chr.19: 35860945-35860945 on Build GRCh38
rs568440666 Chr.19: 35861014-35861014 on Build GRCh38
rs1056760746 Chr.19: 35861071-35861071 on Build GRCh38
rs200047199 Chr.19: 35861077-35861077 on Build GRCh38
rs775695078 Chr.19: 35861067-35861067 on Build GRCh38
rs73928338 Chr.19: 35861004-35861004 on Build GRCh38
rs570978546 Chr.19: 35860978-35860978 on Build GRCh38

View Full Product Details