Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 177230905-177231142 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NFE2L2
Entrez Gene ID
4780
Gene Name
nuclear factor, erythroid 2 like 2
Gene Aliases
HEBP1, NRF2
UniGene
Hs.744006
Gene Chromosome Location
Chr.2: 177230303-177265131 on Build GRCh38
Transcripts
NM_001145412.3 NM_001145413.3 NM_001313900.1 NM_001313901.1 NM_001313902.1 NM_001313903.1 NM_001313904.1 NM_006164.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4982627 c.1589A>T p.D530V Chr.2: 177231014-177231014 on Build GRCh38
6909966 c.1588G>C p.D530H Chr.2: 177231015-177231015 on Build GRCh38
1009926 c.1495C>T p.R499W Chr.2: 177231108-177231108 on Build GRCh38
262393 c.1532C>T p.A511V Chr.2: 177231071-177231071 on Build GRCh38
6984810 c.1636A>C p.N546H Chr.2: 177230967-177230967 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs762320240 Chr.2: 177231012-177231012 on Build GRCh38
rs543741573 Chr.2: 177231027-177231027 on Build GRCh38
rs770583377 Chr.2: 177231050-177231053 on Build GRCh38
rs765842312 Chr.2: 177230957-177230957 on Build GRCh38
rs756129717 Chr.2: 177231116-177231116 on Build GRCh38
rs376346810 Chr.2: 177231001-177231001 on Build GRCh38
rs763001776 Chr.2: 177231073-177231073 on Build GRCh38
rs914770655 Chr.2: 177231099-177231099 on Build GRCh38
rs755639976 Chr.2: 177230935-177230935 on Build GRCh38
rs951842587 Chr.2: 177230934-177230934 on Build GRCh38
rs766602407 Chr.2: 177231074-177231074 on Build GRCh38
rs773337752 Chr.2: 177231040-177231040 on Build GRCh38
rs984737049 Chr.2: 177230947-177230947 on Build GRCh38
rs777072272 Chr.2: 177230995-177230995 on Build GRCh38
rs1045829171 Chr.2: 177231022-177231022 on Build GRCh38
rs376480037 Chr.2: 177231104-177231104 on Build GRCh38
rs1057044 Chr.2: 177231053-177231053 on Build GRCh38
rs1052784015 Chr.2: 177231050-177231050 on Build GRCh38
rs747365318 Chr.2: 177230971-177230971 on Build GRCh38
rs369998189 Chr.2: 177231106-177231106 on Build GRCh38
rs564143578 Chr.2: 177230956-177230956 on Build GRCh38
rs774478415 Chr.2: 177231105-177231105 on Build GRCh38
rs777361176 Chr.2: 177230937-177230937 on Build GRCh38
rs928413267 Chr.2: 177231014-177231014 on Build GRCh38
rs981475836 Chr.2: 177231010-177231010 on Build GRCh38
rs1001449473 Chr.2: 177231039-177231039 on Build GRCh38
rs778477145 Chr.2: 177230967-177230967 on Build GRCh38
rs905868340 Chr.2: 177231035-177231035 on Build GRCh38
rs531558226 Chr.2: 177231019-177231019 on Build GRCh38
rs769038613 Chr.2: 177230973-177230973 on Build GRCh38
rs752627827 Chr.2: 177231107-177231107 on Build GRCh38
rs748812700 Chr.2: 177230944-177230944 on Build GRCh38

View Full Product Details