Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 99333720-99333993 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LRRC28
Entrez Gene ID
123355
Gene Name
leucine rich repeat containing 28
Gene Aliases
-
UniGene
Hs.578684
Gene Chromosome Location
Chr.15: 99251362-99390732 on Build GRCh38
Transcripts
NM_001284400.2 NM_001321675.1 NM_001321676.1 NM_001321677.1 NM_001321678.1 NM_001321679.1 NM_001321680.1 NM_144598.4 NR_135753.1 NR_135754.1 NR_135755.1 NR_135756.1 NR_135757.1 NR_135758.1 NR_135759.1 XM_006720389.3 XM_011521218.2 XM_011521220.1 XM_011521221.2 XM_017021914.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5178762 c.447A>G p.L149L Chr.15: 99333984-99333984 on Build GRCh38
2141639 c.450C>T p.P150P Chr.15: 99333987-99333987 on Build GRCh38
5820907 c.417A>T p.L139L Chr.15: 99333954-99333954 on Build GRCh38
312609 c.451G>A p.E151K Chr.15: 99333988-99333988 on Build GRCh38
434586 c.418G>T p.D140Y Chr.15: 99333955-99333955 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs141689801 Chr.15: 99333926-99333926 on Build GRCh38
rs770423286 Chr.15: 99333965-99333965 on Build GRCh38
rs148635767 Chr.15: 99333903-99333903 on Build GRCh38
rs750690007 Chr.15: 99333915-99333915 on Build GRCh38
rs1039044230 Chr.15: 99333768-99333768 on Build GRCh38
rs753768597 Chr.15: 99333900-99333900 on Build GRCh38
rs888153132 Chr.15: 99333820-99333820 on Build GRCh38
rs771749227 Chr.15: 99333931-99333931 on Build GRCh38
rs959669374 Chr.15: 99333968-99333968 on Build GRCh38
rs929773044 Chr.15: 99333778-99333778 on Build GRCh38
rs377428226 Chr.15: 99333884-99333884 on Build GRCh38
rs777400728 Chr.15: 99333951-99333951 on Build GRCh38
rs754088714 Chr.15: 99333925-99333925 on Build GRCh38
rs780502964 Chr.15: 99333919-99333919 on Build GRCh38
rs756276353 Chr.15: 99333918-99333918 on Build GRCh38
rs186458223 Chr.15: 99333861-99333861 on Build GRCh38
rs376380327 Chr.15: 99333863-99333863 on Build GRCh38
rs539526677 Chr.15: 99333909-99333909 on Build GRCh38
rs941010088 Chr.15: 99333878-99333878 on Build GRCh38
rs371208978 Chr.15: 99333894-99333894 on Build GRCh38
rs746681650 Chr.15: 99333959-99333959 on Build GRCh38
rs891697092 Chr.15: 99333833-99333833 on Build GRCh38
rs1047165425 Chr.15: 99333803-99333803 on Build GRCh38
rs900477486 Chr.15: 99333780-99333780 on Build GRCh38
rs1010509211 Chr.15: 99333858-99333858 on Build GRCh38
rs779348807 Chr.15: 99333930-99333930 on Build GRCh38
rs757188580 Chr.15: 99333970-99333970 on Build GRCh38

View Full Product Details