Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 30911012-30911251 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GTF2H4
Entrez Gene ID
2968
Gene Name
general transcription factor IIH subunit 4
Gene Aliases
P52, TFB2, TFIIH
UniGene
Hs.485070
Gene Chromosome Location
Chr.6: 30908200-30914103 on Build GRCh38
Transcripts
NM_001517.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6499174 c.636C>G p.L212L Chr.6: 30911233-30911233 on Build GRCh38
1077480 c.628G>A p.A210T Chr.6: 30911225-30911225 on Build GRCh38
3252363 c.622A>C p.T208P Chr.6: 30911219-30911219 on Build GRCh38
4689161 c.578C>T p.P193L Chr.6: 30911175-30911175 on Build GRCh38
6386670 c.626C>A p.P209Q Chr.6: 30911223-30911223 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs1055928137 Chr.6: 30911082-30911082 on Build GRCh38
rs761913714 Chr.6: 30911223-30911223 on Build GRCh38
rs546171158 Chr.6: 30911109-30911109 on Build GRCh38
rs947504510 Chr.6: 30911101-30911101 on Build GRCh38
rs753617665 Chr.6: 30911225-30911225 on Build GRCh38
rs1041776789 Chr.6: 30911114-30911114 on Build GRCh38
rs371323226 Chr.6: 30911149-30911149 on Build GRCh38
rs746591176 Chr.6: 30911180-30911180 on Build GRCh38
rs138478436 Chr.6: 30911219-30911219 on Build GRCh38
rs111368171 Chr.6: 30911176-30911176 on Build GRCh38
rs914724889 Chr.6: 30911088-30911088 on Build GRCh38
rs374908109 Chr.6: 30911175-30911175 on Build GRCh38
rs377712535 Chr.6: 30911209-30911209 on Build GRCh38
rs2074512 Chr.6: 30911142-30911142 on Build GRCh38
rs902139026 Chr.6: 30911139-30911139 on Build GRCh38
rs778452455 Chr.6: 30911168-30911168 on Build GRCh38
rs766067957 Chr.6: 30911158-30911158 on Build GRCh38
rs765445291 Chr.6: 30911120-30911120 on Build GRCh38
rs754907802 Chr.6: 30911165-30911165 on Build GRCh38
rs150834984 Chr.6: 30911191-30911191 on Build GRCh38
rs1020639460 Chr.6: 30911087-30911087 on Build GRCh38
rs3218816 Chr.6: 30911080-30911080 on Build GRCh38
rs753854558 Chr.6: 30911160-30911160 on Build GRCh38
rs775703945 Chr.6: 30911128-30911128 on Build GRCh38
rs764033929 Chr.6: 30911213-30911213 on Build GRCh38
rs928851726 Chr.6: 30911050-30911050 on Build GRCh38
rs770408294 Chr.6: 30911192-30911192 on Build GRCh38
rs770431577 Chr.6: 30911066-30911066 on Build GRCh38
rs143011675 Chr.6: 30911224-30911224 on Build GRCh38
rs771374538 Chr.6: 30911183-30911183 on Build GRCh38
rs141405959 Chr.6: 30911188-30911188 on Build GRCh38
rs763321470 Chr.6: 30911132-30911132 on Build GRCh38
rs34535454 Chr.6: 30911190-30911190 on Build GRCh38
rs760094679 Chr.6: 30911118-30911118 on Build GRCh38
rs1054903715 Chr.6: 30911064-30911064 on Build GRCh38
rs777216519 Chr.6: 30911110-30911110 on Build GRCh38

View Full Product Details