Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 41378909-41379170 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KRT34
Entrez Gene ID
3885
Gene Name
keratin 34
Gene Aliases
HA4, Ha-4, KRTHA4, hHa4
UniGene
Hs.296942
Gene Chromosome Location
Chr.17: 41377650-41383816 on Build GRCh38
Transcripts
NM_021013.3 XM_011524793.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2149202 c.1086C>T p.N362N Chr.17: 41379093-41379093 on Build GRCh38
212909 c.1174G>C p.E392Q Chr.17: 41379005-41379005 on Build GRCh38
3517314 c.1111C>T p.R371C Chr.17: 41379068-41379068 on Build GRCh38
3517313 c.1216G>A p.D406N Chr.17: 41378963-41378963 on Build GRCh38
1686256 c.1169G>A p.R390Q Chr.17: 41379010-41379010 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs572705563 Chr.17: 41379138-41379138 on Build GRCh38
rs753762289 Chr.17: 41379024-41379024 on Build GRCh38
rs2071599 Chr.17: 41379136-41379136 on Build GRCh38
rs775908971 Chr.17: 41379106-41379106 on Build GRCh38
rs61740668 Chr.17: 41379053-41379053 on Build GRCh38
rs753797165 Chr.17: 41378966-41378966 on Build GRCh38
rs765551296 Chr.17: 41379140-41379140 on Build GRCh38
rs765270303 Chr.17: 41379075-41379075 on Build GRCh38
rs755328052 Chr.17: 41379099-41379099 on Build GRCh38
rs748114147 Chr.17: 41378989-41378989 on Build GRCh38
rs779449926 Chr.17: 41379100-41379100 on Build GRCh38
rs762043346 Chr.17: 41379084-41379084 on Build GRCh38
rs866482029 Chr.17: 41379149-41379149 on Build GRCh38
rs759197812 Chr.17: 41379071-41379071 on Build GRCh38
rs201791417 Chr.17: 41379108-41379108 on Build GRCh38
rs759553168 Chr.17: 41379027-41379027 on Build GRCh38
rs142124448 Chr.17: 41379143-41379143 on Build GRCh38
rs747720099 Chr.17: 41379057-41379057 on Build GRCh38
rs769700932 Chr.17: 41379120-41379120 on Build GRCh38
rs574453106 Chr.17: 41379009-41379009 on Build GRCh38
rs756379610 Chr.17: 41379093-41379093 on Build GRCh38
rs556713103 Chr.17: 41379067-41379067 on Build GRCh38
rs148874673 Chr.17: 41379150-41379150 on Build GRCh38
rs769002695 Chr.17: 41379005-41379005 on Build GRCh38
rs746536930 Chr.17: 41379147-41379147 on Build GRCh38
rs771110114 Chr.17: 41379045-41379045 on Build GRCh38
rs370544169 Chr.17: 41379092-41379092 on Build GRCh38
rs760204033 Chr.17: 41378979-41378979 on Build GRCh38
rs751651630 Chr.17: 41378983-41378983 on Build GRCh38
rs750645909 Chr.17: 41379044-41379044 on Build GRCh38
rs771555272 Chr.17: 41379059-41379059 on Build GRCh38
rs764072288 Chr.17: 41379123-41379123 on Build GRCh38
rs1049965115 Chr.17: 41379148-41379148 on Build GRCh38
rs200529690 Chr.17: 41379046-41379046 on Build GRCh38
rs775563371 Chr.17: 41379082-41379082 on Build GRCh38
rs139425895 Chr.17: 41379036-41379036 on Build GRCh38
rs866220230 Chr.17: 41379133-41379133 on Build GRCh38
rs750229246 Chr.17: 41378952-41378952 on Build GRCh38
rs183229015 Chr.17: 41379139-41379139 on Build GRCh38
rs141543111 Chr.17: 41379021-41379021 on Build GRCh38
rs767106246 Chr.17: 41378947-41378947 on Build GRCh38
rs529354447 Chr.17: 41379068-41379068 on Build GRCh38
rs780484325 Chr.17: 41379003-41379003 on Build GRCh38
rs749770713 Chr.17: 41379004-41379004 on Build GRCh38
rs780496865 Chr.17: 41379051-41379051 on Build GRCh38
rs745725606 Chr.17: 41379114-41379114 on Build GRCh38
rs117649171 Chr.17: 41378964-41378964 on Build GRCh38
rs568335050 Chr.17: 41379060-41379060 on Build GRCh38
rs773354255 Chr.17: 41379128-41379128 on Build GRCh38
rs200232759 Chr.17: 41379011-41379011 on Build GRCh38
rs144999065 Chr.17: 41379040-41379040 on Build GRCh38
rs148645199 Chr.17: 41379113-41379113 on Build GRCh38
rs61740667 Chr.17: 41379032-41379032 on Build GRCh38
rs35285640 Chr.17: 41379065-41379066 on Build GRCh38
rs754233528 Chr.17: 41379097-41379097 on Build GRCh38
rs144311709 Chr.17: 41379102-41379102 on Build GRCh38
rs779046810 Chr.17: 41378988-41378988 on Build GRCh38
rs61740669 Chr.17: 41379065-41379065 on Build GRCh38
rs754958901 Chr.17: 41378984-41378984 on Build GRCh38
rs546536544 Chr.17: 41379056-41379056 on Build GRCh38
rs770634433 Chr.17: 41379110-41379110 on Build GRCh38
rs1053377458 Chr.17: 41379001-41379001 on Build GRCh38
rs1043572213 Chr.17: 41379117-41379117 on Build GRCh38
rs749566121 Chr.17: 41379052-41379052 on Build GRCh38
rs374681338 Chr.17: 41379016-41379016 on Build GRCh38
rs148769355 Chr.17: 41379020-41379020 on Build GRCh38
rs760449691 Chr.17: 41378963-41378963 on Build GRCh38
rs773873539 Chr.17: 41379007-41379007 on Build GRCh38
rs199674249 Chr.17: 41379017-41379017 on Build GRCh38
rs766453950 Chr.17: 41379094-41379094 on Build GRCh38
rs775618483 Chr.17: 41379122-41379122 on Build GRCh38
rs908234530 Chr.17: 41378960-41378960 on Build GRCh38
rs758460469 Chr.17: 41378974-41378974 on Build GRCh38
rs760604152 Chr.17: 41379129-41379129 on Build GRCh38
rs36019136 Chr.17: 41379125-41379125 on Build GRCh38
rs916008571 Chr.17: 41378940-41378940 on Build GRCh38
rs200168141 Chr.17: 41379105-41379105 on Build GRCh38
rs777972734 Chr.17: 41378982-41378982 on Build GRCh38
rs145538860 Chr.17: 41379144-41379144 on Build GRCh38

View Full Product Details