Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 54281301-54281574 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HNRNPA1
Entrez Gene ID
3178
Gene Name
heterogeneous nuclear ribonucleoprotein A1
Gene Aliases
ALS19, ALS20, HNRPA1, HNRPA1L3, IBMPFD3, UP 1, hnRNP A1, hnRNP-A1
UniGene
Hs.546261
Gene Chromosome Location
Chr.12: 54280690-54287088 on Build GRCh38
Transcripts
NM_002136.3 NM_031157.3 NR_135167.1 XM_005268826.1 XM_017019251.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3722077 c.70G>T p.E24* Chr.12: 54281440-54281440 on Build GRCh38
6072886 c.20C>G p.P7R Chr.12: 54281390-54281390 on Build GRCh38
3722075 c.79G>T p.D27Y Chr.12: 54281449-54281449 on Build GRCh38
4444421 c.55G>A p.G19R Chr.12: 54281425-54281425 on Build GRCh38
123628 c.124G>C p.D42H Chr.12: 54281494-54281494 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs755744884 Chr.12: 54281372-54281373 on Build GRCh38
rs775845623 Chr.12: 54281517-54281517 on Build GRCh38
rs941085168 Chr.12: 54281551-54281551 on Build GRCh38
rs749587445 Chr.12: 54281518-54281518 on Build GRCh38
rs753279252 Chr.12: 54281536-54281536 on Build GRCh38
rs767688009 Chr.12: 54281381-54281381 on Build GRCh38
rs755921419 Chr.12: 54281343-54281343 on Build GRCh38
rs371472630 Chr.12: 54281514-54281514 on Build GRCh38
rs781407881 Chr.12: 54281337-54281337 on Build GRCh38
rs778592922 Chr.12: 54281352-54281352 on Build GRCh38
rs754983468 Chr.12: 54281416-54281416 on Build GRCh38
rs941369770 Chr.12: 54281328-54281328 on Build GRCh38
rs1049513476 Chr.12: 54281545-54281545 on Build GRCh38
rs780816260 Chr.12: 54281553-54281553 on Build GRCh38
rs769216459 Chr.12: 54281520-54281520 on Build GRCh38
rs2071391 Chr.12: 54281329-54281329 on Build GRCh38
rs761874206 Chr.12: 54281378-54281378 on Build GRCh38
rs747607504 Chr.12: 54281356-54281356 on Build GRCh38
rs777373779 Chr.12: 54281543-54281543 on Build GRCh38
rs560406331 Chr.12: 54281548-54281548 on Build GRCh38
rs773121968 Chr.12: 54281526-54281526 on Build GRCh38
rs899452646 Chr.12: 54281333-54281333 on Build GRCh38
rs537918449 Chr.12: 54281341-54281341 on Build GRCh38
rs373601918 Chr.12: 54281362-54281362 on Build GRCh38
rs755898648 Chr.12: 54281388-54281388 on Build GRCh38
rs757914524 Chr.12: 54281463-54281463 on Build GRCh38
rs190679271 Chr.12: 54281536-54281536 on Build GRCh38
rs754922339 Chr.12: 54281351-54281351 on Build GRCh38
rs746278274 Chr.12: 54281364-54281364 on Build GRCh38
rs373496287 Chr.12: 54281549-54281549 on Build GRCh38
rs201465178 Chr.12: 54281368-54281368 on Build GRCh38
rs28758594 Chr.12: 54281478-54281478 on Build GRCh38
rs994135955 Chr.12: 54281344-54281344 on Build GRCh38
rs761844470 Chr.12: 54281522-54281522 on Build GRCh38
rs749204083 Chr.12: 54281346-54281346 on Build GRCh38
rs777932166 Chr.12: 54281350-54281351 on Build GRCh38
rs778930233 Chr.12: 54281427-54281427 on Build GRCh38
rs916151919 Chr.12: 54281332-54281332 on Build GRCh38
rs753695201 Chr.12: 54281532-54281532 on Build GRCh38
rs746642933 Chr.12: 54281510-54281510 on Build GRCh38
rs754818936 Chr.12: 54281533-54281533 on Build GRCh38
rs747731298 Chr.12: 54281460-54281460 on Build GRCh38
rs750633785 Chr.12: 54281383-54281383 on Build GRCh38
rs766224014 Chr.12: 54281390-54281390 on Build GRCh38
rs61910737 Chr.12: 54281494-54281494 on Build GRCh38
rs752660681 Chr.12: 54281537-54281537 on Build GRCh38
rs186328944 Chr.12: 54281535-54281535 on Build GRCh38
rs925777534 Chr.12: 54281448-54281448 on Build GRCh38
rs757483485 Chr.12: 54281335-54281335 on Build GRCh38
rs372935253 Chr.12: 54281363-54281363 on Build GRCh38
rs1011227682 Chr.12: 54281496-54281496 on Build GRCh38
rs376518306 Chr.12: 54281371-54281371 on Build GRCh38
rs199689675 Chr.12: 54281521-54281521 on Build GRCh38
rs767599936 Chr.12: 54281525-54281525 on Build GRCh38
rs757887840 Chr.12: 54281538-54281538 on Build GRCh38
rs753643060 Chr.12: 54281400-54281400 on Build GRCh38
rs777342318 Chr.12: 54281490-54281490 on Build GRCh38
rs777323931 Chr.12: 54281529-54281529 on Build GRCh38
rs774417100 Chr.12: 54281374-54281374 on Build GRCh38
rs201410105 Chr.12: 54281373-54281373 on Build GRCh38
rs766458005 Chr.12: 54281527-54281527 on Build GRCh38

View Full Product Details