Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 88814437-88814710 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LIPM
Entrez Gene ID
340654
Gene Name
lipase family member M
Gene Aliases
LIPL3, bA304I5.1
UniGene
Hs.55118
Gene Chromosome Location
Chr.10: 88802730-88820857 on Build GRCh38
Transcripts
NM_001128215.1 XM_011539748.2 XM_011539751.2 XM_011539752.2 XM_011539753.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3981181 c.538A>T p.I180F Chr.10: 88814603-88814603 on Build GRCh38
6262772 c.345-4G>T p.? Chr.10: 88814526-88814526 on Build GRCh38
6262771 c.465-4G>T p.? Chr.10: 88814526-88814526 on Build GRCh38
7375122 c.428T>A p.V143D Chr.10: 88814613-88814613 on Build GRCh38
6293419 c.397C>A p.Q133K Chr.10: 88814582-88814582 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs759244129 Chr.10: 88814672-88814672 on Build GRCh38
rs908455199 Chr.10: 88814683-88814683 on Build GRCh38
rs765950068 Chr.10: 88814533-88814533 on Build GRCh38
rs303524 Chr.10: 88814465-88814465 on Build GRCh38
rs964438506 Chr.10: 88814612-88814612 on Build GRCh38
rs947268069 Chr.10: 88814622-88814631 on Build GRCh38
rs570098337 Chr.10: 88814482-88814482 on Build GRCh38
rs769750406 Chr.10: 88814614-88814614 on Build GRCh38
rs751873207 Chr.10: 88814592-88814592 on Build GRCh38
rs765368181 Chr.10: 88814533-88814533 on Build GRCh38
rs770530196 Chr.10: 88814644-88814644 on Build GRCh38
rs750439349 Chr.10: 88814558-88814558 on Build GRCh38
rs200508607 Chr.10: 88814590-88814590 on Build GRCh38
rs371475612 Chr.10: 88814607-88814607 on Build GRCh38
rs558938025 Chr.10: 88814643-88814643 on Build GRCh38
rs758555172 Chr.10: 88814572-88814572 on Build GRCh38
rs764560934 Chr.10: 88814480-88814480 on Build GRCh38
rs1004040373 Chr.10: 88814589-88814589 on Build GRCh38
rs773811351 Chr.10: 88814668-88814668 on Build GRCh38
rs781687109 Chr.10: 88814521-88814521 on Build GRCh38
rs777723800 Chr.10: 88814629-88814629 on Build GRCh38
rs756271856 Chr.10: 88814602-88814602 on Build GRCh38
rs546873976 Chr.10: 88814615-88814615 on Build GRCh38
rs906875298 Chr.10: 88814503-88814503 on Build GRCh38
rs915035129 Chr.10: 88814666-88814666 on Build GRCh38
rs982771678 Chr.10: 88814681-88814681 on Build GRCh38
rs1029737653 Chr.10: 88814632-88814632 on Build GRCh38
rs1031253984 Chr.10: 88814550-88814550 on Build GRCh38
rs762435800 Chr.10: 88814487-88814487 on Build GRCh38
rs1011476607 Chr.10: 88814591-88814591 on Build GRCh38
rs1027766437 Chr.10: 88814496-88814496 on Build GRCh38
rs948516095 Chr.10: 88814471-88814471 on Build GRCh38
rs754699630 Chr.10: 88814597-88814597 on Build GRCh38
rs113548543 Chr.10: 88814465-88814465 on Build GRCh38

View Full Product Details