Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 6862423-6862694 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KIAA0232
Entrez Gene ID
9778
Gene Name
KIAA0232
Gene Aliases
-
UniGene
Hs.79276
Gene Chromosome Location
Chr.4: 6782732-6884172 on Build GRCh38
Transcripts
NM_001100590.1 NM_014743.2 XM_005248041.4 XM_011513598.2 XM_017008860.1 XM_017008861.1 XM_017008862.1 XM_017008863.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5533210 c.2249C>T p.S750L Chr.4: 6862631-6862631 on Build GRCh38
4927529 c.2090T>A p.F697Y Chr.4: 6862472-6862472 on Build GRCh38
7263552 c.2248T>G p.S750A Chr.4: 6862630-6862630 on Build GRCh38
7201264 c.2154A>C p.E718D Chr.4: 6862536-6862536 on Build GRCh38
7228038 c.2076C>A p.T692T Chr.4: 6862458-6862458 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs773901529 Chr.4: 6862568-6862568 on Build GRCh38
rs755080807 Chr.4: 6862600-6862600 on Build GRCh38
rs746251464 Chr.4: 6862453-6862453 on Build GRCh38
rs753974837 Chr.4: 6862479-6862479 on Build GRCh38
rs201903691 Chr.4: 6862632-6862632 on Build GRCh38
rs747343639 Chr.4: 6862533-6862533 on Build GRCh38
rs767048158 Chr.4: 6862584-6862584 on Build GRCh38
rs1034400334 Chr.4: 6862536-6862536 on Build GRCh38
rs371066886 Chr.4: 6862485-6862485 on Build GRCh38
rs570786567 Chr.4: 6862648-6862648 on Build GRCh38
rs748361911 Chr.4: 6862486-6862486 on Build GRCh38
rs200375350 Chr.4: 6862538-6862538 on Build GRCh38
rs752693848 Chr.4: 6862631-6862631 on Build GRCh38
rs777850900 Chr.4: 6862644-6862644 on Build GRCh38
rs761576076 Chr.4: 6862575-6862575 on Build GRCh38
rs990589245 Chr.4: 6862629-6862629 on Build GRCh38
rs879442534 Chr.4: 6862515-6862515 on Build GRCh38
rs760447432 Chr.4: 6862596-6862596 on Build GRCh38
rs183638611 Chr.4: 6862521-6862521 on Build GRCh38
rs758707395 Chr.4: 6862493-6862493 on Build GRCh38
rs765308181 Chr.4: 6862628-6862628 on Build GRCh38
rs374089474 Chr.4: 6862564-6862564 on Build GRCh38
rs766316465 Chr.4: 6862597-6862597 on Build GRCh38
rs777222469 Chr.4: 6862552-6862552 on Build GRCh38
rs748815244 Chr.4: 6862556-6862556 on Build GRCh38
rs754991049 Chr.4: 6862481-6862481 on Build GRCh38
rs1002972390 Chr.4: 6862495-6862495 on Build GRCh38
rs773128574 Chr.4: 6862592-6862592 on Build GRCh38
rs140804894 Chr.4: 6862599-6862599 on Build GRCh38
rs746331960 Chr.4: 6862662-6862662 on Build GRCh38
rs766302050 Chr.4: 6862457-6862457 on Build GRCh38
rs750608465 Chr.4: 6862621-6862621 on Build GRCh38
rs376091389 Chr.4: 6862646-6862646 on Build GRCh38
rs747392975 Chr.4: 6862645-6862645 on Build GRCh38

View Full Product Details