Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 8891385-8891610 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MFHAS1
Entrez Gene ID
9258
Gene Name
malignant fibrous histiocytoma amplified sequence 1
Gene Aliases
LRRC65, MASL1
UniGene
Hs.379414
Gene Chromosome Location
Chr.8: 8783354-8893621 on Build GRCh38
Transcripts
NM_004225.2 XM_011543852.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7270160 c.1546G>A p.V516M Chr.8: 8891513-8891513 on Build GRCh38
2789731 c.1494C>T p.Y498Y Chr.8: 8891565-8891565 on Build GRCh38
2789730 c.1529G>A p.R510H Chr.8: 8891530-8891530 on Build GRCh38
3651505 c.1452T>G p.Y484* Chr.8: 8891607-8891607 on Build GRCh38
4138758 c.1625G>T p.C542F Chr.8: 8891434-8891434 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs760052526 Chr.8: 8891407-8891407 on Build GRCh38
rs766378290 Chr.8: 8891579-8891579 on Build GRCh38
rs1012974305 Chr.8: 8891492-8891492 on Build GRCh38
rs779997040 Chr.8: 8891520-8891520 on Build GRCh38
rs747806262 Chr.8: 8891480-8891480 on Build GRCh38
rs773400094 Chr.8: 8891570-8891570 on Build GRCh38
rs149204523 Chr.8: 8891485-8891485 on Build GRCh38
rs387261 Chr.8: 8891420-8891420 on Build GRCh38
rs760221622 Chr.8: 8891562-8891562 on Build GRCh38
rs908398765 Chr.8: 8891555-8891555 on Build GRCh38
rs763484243 Chr.8: 8891426-8891426 on Build GRCh38
rs765808857 Chr.8: 8891500-8891502 on Build GRCh38
rs565604644 Chr.8: 8891445-8891445 on Build GRCh38
rs771755259 Chr.8: 8891494-8891494 on Build GRCh38
rs764426249 Chr.8: 8891439-8891439 on Build GRCh38
rs558634917 Chr.8: 8891473-8891473 on Build GRCh38
rs747787696 Chr.8: 8891533-8891533 on Build GRCh38
rs769006332 Chr.8: 8891558-8891558 on Build GRCh38
rs749143961 Chr.8: 8891474-8891474 on Build GRCh38
rs529772352 Chr.8: 8891583-8891583 on Build GRCh38
rs777057964 Chr.8: 8891405-8891405 on Build GRCh38
rs372634552 Chr.8: 8891443-8891443 on Build GRCh38
rs142063688 Chr.8: 8891538-8891538 on Build GRCh38
rs148315756 Chr.8: 8891531-8891531 on Build GRCh38
rs405251 Chr.8: 8891421-8891421 on Build GRCh38
rs772768467 Chr.8: 8891486-8891486 on Build GRCh38
rs770370271 Chr.8: 8891541-8891541 on Build GRCh38
rs202143016 Chr.8: 8891539-8891539 on Build GRCh38
rs754357622 Chr.8: 8891561-8891562 on Build GRCh38
rs1033009644 Chr.8: 8891476-8891476 on Build GRCh38
rs781423982 Chr.8: 8891465-8891465 on Build GRCh38
rs760800087 Chr.8: 8891571-8891571 on Build GRCh38
rs763372213 Chr.8: 8891557-8891557 on Build GRCh38
rs764396180 Chr.8: 8891507-8891507 on Build GRCh38
rs879315752 Chr.8: 8891420-8891421 on Build GRCh38
rs370204052 Chr.8: 8891408-8891408 on Build GRCh38
rs200309914 Chr.8: 8891567-8891567 on Build GRCh38
rs763326843 Chr.8: 8891506-8891506 on Build GRCh38
rs754931910 Chr.8: 8891585-8891585 on Build GRCh38
rs751736035 Chr.8: 8891508-8891508 on Build GRCh38
rs1043571879 Chr.8: 8891529-8891529 on Build GRCh38
rs765573852 Chr.8: 8891410-8891410 on Build GRCh38
rs760032458 Chr.8: 8891489-8891489 on Build GRCh38
rs750325866 Chr.8: 8891466-8891466 on Build GRCh38
rs143346462 Chr.8: 8891514-8891514 on Build GRCh38
rs138267533 Chr.8: 8891478-8891478 on Build GRCh38
rs560880992 Chr.8: 8891534-8891534 on Build GRCh38
rs776024190 Chr.8: 8891544-8891544 on Build GRCh38
rs753754133 Chr.8: 8891522-8891522 on Build GRCh38
rs767689063 Chr.8: 8891509-8891509 on Build GRCh38

View Full Product Details