Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 204410326-204410553 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PPP1R15B
Entrez Gene ID
84919
Gene Name
protein phosphatase 1 regulatory subunit 15B
Gene Aliases
CREP, MSSGM2
UniGene
Hs.304376
Gene Chromosome Location
Chr.1: 204398684-204411880 on Build GRCh38
Transcripts
NM_032833.4 XM_005245551.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7256342 c.997C>A p.R333R Chr.1: 204410415-204410415 on Build GRCh38
6346326 c.972C>A p.S324R Chr.1: 204410440-204410440 on Build GRCh38
6459943 c.1078T>A p.S360T Chr.1: 204410334-204410334 on Build GRCh38
1338073 c.1048G>A p.A350T Chr.1: 204410364-204410364 on Build GRCh38
6251826 c.920C>A p.A307D Chr.1: 204410492-204410492 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs762013594 Chr.1: 204410502-204410502 on Build GRCh38
rs546031521 Chr.1: 204410443-204410443 on Build GRCh38
rs775288516 Chr.1: 204410497-204410497 on Build GRCh38
rs751024564 Chr.1: 204410452-204410454 on Build GRCh38
rs988501583 Chr.1: 204410490-204410490 on Build GRCh38
rs778301648 Chr.1: 204410462-204410462 on Build GRCh38
rs765916830 Chr.1: 204410429-204410446 on Build GRCh38
rs767849759 Chr.1: 204410425-204410425 on Build GRCh38
rs913430361 Chr.1: 204410527-204410527 on Build GRCh38
rs144269029 Chr.1: 204410472-204410472 on Build GRCh38
rs750600395 Chr.1: 204410428-204410428 on Build GRCh38
rs994252931 Chr.1: 204410386-204410386 on Build GRCh38
rs767477366 Chr.1: 204410510-204410510 on Build GRCh38
rs754374161 Chr.1: 204410446-204410446 on Build GRCh38
rs1018840318 Chr.1: 204410449-204410449 on Build GRCh38
rs932050075 Chr.1: 204410468-204410468 on Build GRCh38
rs769703494 Chr.1: 204410494-204410494 on Build GRCh38
rs761948500 Chr.1: 204410423-204410423 on Build GRCh38
rs564562381 Chr.1: 204410390-204410390 on Build GRCh38
rs368649349 Chr.1: 204410352-204410352 on Build GRCh38
rs558337906 Chr.1: 204410457-204410457 on Build GRCh38
rs766752711 Chr.1: 204410528-204410528 on Build GRCh38
rs776515101 Chr.1: 204410487-204410487 on Build GRCh38
rs11799830 Chr.1: 204410404-204410404 on Build GRCh38
rs777530382 Chr.1: 204410471-204410471 on Build GRCh38
rs756523811 Chr.1: 204410432-204410432 on Build GRCh38
rs751597239 Chr.1: 204410422-204410422 on Build GRCh38
rs773467228 Chr.1: 204410521-204410521 on Build GRCh38
rs747780003 Chr.1: 204410370-204410370 on Build GRCh38
rs778447234 Chr.1: 204410362-204410362 on Build GRCh38
rs755430265 Chr.1: 204410459-204410459 on Build GRCh38
rs757832337 Chr.1: 204410470-204410470 on Build GRCh38
rs771699532 Chr.1: 204410377-204410377 on Build GRCh38
rs749777475 Chr.1: 204410348-204410348 on Build GRCh38
rs3014626 Chr.1: 204410489-204410489 on Build GRCh38
rs770876375 Chr.1: 204410482-204410482 on Build GRCh38
rs760847763 Chr.1: 204410525-204410525 on Build GRCh38
rs898545713 Chr.1: 204410376-204410376 on Build GRCh38
rs772734091 Chr.1: 204410378-204410378 on Build GRCh38
rs764288783 Chr.1: 204410409-204410409 on Build GRCh38
rs773569524 Chr.1: 204410363-204410363 on Build GRCh38
rs1042018791 Chr.1: 204410518-204410518 on Build GRCh38
rs866458892 Chr.1: 204410410-204410410 on Build GRCh38
rs747758689 Chr.1: 204410469-204410469 on Build GRCh38
rs776745598 Chr.1: 204410403-204410403 on Build GRCh38
rs760603744 Chr.1: 204410382-204410382 on Build GRCh38

View Full Product Details