Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 69844142-69844367 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GMCL1
Entrez Gene ID
64395
Gene Name
germ cell-less, spermatogenesis associated 1
Gene Aliases
BTBD13, GCL, GCL1, SPATA29
UniGene
Hs.293971
Gene Chromosome Location
Chr.2: 69829606-69881396 on Build GRCh38
Transcripts
NM_178439.4 XM_011533033.1 XM_011533034.2 XM_017004704.1 XM_017004705.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7164457 c.727C>T p.H243Y Chr.2: 69844165-69844165 on Build GRCh38
4417013 c.735T>C p.N245N Chr.2: 69844173-69844173 on Build GRCh38
6671955 c.746T>G p.F249C Chr.2: 69844184-69844184 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs1007671801 Chr.2: 69844327-69844327 on Build GRCh38
rs988065405 Chr.2: 69844170-69844170 on Build GRCh38
rs757085207 Chr.2: 69844178-69844178 on Build GRCh38
rs1016661448 Chr.2: 69844324-69844324 on Build GRCh38
rs368833752 Chr.2: 69844202-69844202 on Build GRCh38
rs775433589 Chr.2: 69844209-69844209 on Build GRCh38
rs75706731 Chr.2: 69844261-69844261 on Build GRCh38
rs969559124 Chr.2: 69844331-69844331 on Build GRCh38
rs1025307492 Chr.2: 69844264-69844264 on Build GRCh38
rs991745045 Chr.2: 69844243-69844243 on Build GRCh38
rs768197007 Chr.2: 69844216-69844216 on Build GRCh38
rs1048866540 Chr.2: 69844310-69844310 on Build GRCh38
rs774192463 Chr.2: 69844223-69844223 on Build GRCh38
rs149540745 Chr.2: 69844206-69844206 on Build GRCh38
rs548449166 Chr.2: 69844211-69844211 on Build GRCh38
rs527277272 Chr.2: 69844260-69844260 on Build GRCh38
rs752930018 Chr.2: 69844173-69844174 on Build GRCh38
rs771810216 Chr.2: 69844231-69844231 on Build GRCh38
rs918979732 Chr.2: 69844282-69844282 on Build GRCh38
rs35879465 Chr.2: 69844185-69844185 on Build GRCh38
rs200259207 Chr.2: 69844229-69844229 on Build GRCh38
rs781222465 Chr.2: 69844169-69844170 on Build GRCh38
rs781000060 Chr.2: 69844201-69844201 on Build GRCh38
rs886807539 Chr.2: 69844273-69844273 on Build GRCh38
rs957627531 Chr.2: 69844224-69844224 on Build GRCh38
rs916186582 Chr.2: 69844259-69844259 on Build GRCh38
rs570177724 Chr.2: 69844299-69844299 on Build GRCh38
rs773735056 Chr.2: 69844241-69844241 on Build GRCh38
rs532589550 Chr.2: 69844320-69844320 on Build GRCh38
rs34972580 Chr.2: 69844251-69844252 on Build GRCh38
rs17263534 Chr.2: 69844173-69844173 on Build GRCh38

View Full Product Details