Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 64198739-64199012 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MYT1
Entrez Gene ID
4661
Gene Name
myelin transcription factor 1
Gene Aliases
C20orf36, MTF1, MYTI, NZF2, PLPB1, ZC2H2C1, ZC2HC4A
UniGene
Hs.279562
Gene Chromosome Location
Chr.20: 64164474-64242253 on Build GRCh38
Transcripts
NM_004535.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4705267 c.52C>T p.R18* Chr.20: 64198913-64198913 on Build GRCh38
4705263 c.13A>T p.N5Y Chr.20: 64198874-64198874 on Build GRCh38
4705266 c.52C>T p.R18* Chr.20: 64198913-64198913 on Build GRCh38
6526138 c.41C>A p.S14Y Chr.20: 64198902-64198902 on Build GRCh38
6526140 c.41C>A p.S14Y Chr.20: 64198902-64198902 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs762794729 Chr.20: 64198929-64198929 on Build GRCh38
rs574959009 Chr.20: 64198934-64198934 on Build GRCh38
rs954581097 Chr.20: 64198881-64198881 on Build GRCh38
rs202164355 Chr.20: 64198844-64198844 on Build GRCh38
rs370968419 Chr.20: 64198907-64198907 on Build GRCh38
rs753848595 Chr.20: 64198948-64198948 on Build GRCh38
rs761900241 Chr.20: 64198906-64198906 on Build GRCh38
rs771745530 Chr.20: 64198858-64198858 on Build GRCh38
rs2427613 Chr.20: 64198926-64198926 on Build GRCh38
rs367847705 Chr.20: 64198899-64198899 on Build GRCh38
rs757773977 Chr.20: 64198961-64198961 on Build GRCh38
rs916742088 Chr.20: 64198990-64198990 on Build GRCh38
rs377127283 Chr.20: 64198967-64198967 on Build GRCh38
rs777972769 Chr.20: 64198940-64198940 on Build GRCh38
rs755265896 Chr.20: 64198828-64198828 on Build GRCh38
rs952888845 Chr.20: 64198963-64198963 on Build GRCh38
rs919967523 Chr.20: 64198944-64198944 on Build GRCh38
rs200123158 Chr.20: 64198840-64198840 on Build GRCh38
rs2295450 Chr.20: 64198937-64198937 on Build GRCh38
rs781495487 Chr.20: 64198832-64198832 on Build GRCh38
rs779323629 Chr.20: 64198962-64198962 on Build GRCh38
rs777693253 Chr.20: 64198841-64198841 on Build GRCh38
rs201067948 Chr.20: 64198843-64198843 on Build GRCh38
rs767653618 Chr.20: 64198932-64198932 on Build GRCh38
rs373252869 Chr.20: 64198815-64198815 on Build GRCh38
rs772475976 Chr.20: 64198966-64198966 on Build GRCh38
rs375283874 Chr.20: 64198798-64198798 on Build GRCh38
rs1008694011 Chr.20: 64198915-64198915 on Build GRCh38
rs768256568 Chr.20: 64198896-64198896 on Build GRCh38
rs933038383 Chr.20: 64198766-64198766 on Build GRCh38
rs752749174 Chr.20: 64198836-64198836 on Build GRCh38
rs779305121 Chr.20: 64198848-64198848 on Build GRCh38
rs775162910 Chr.20: 64198887-64198887 on Build GRCh38
rs372470070 Chr.20: 64198965-64198965 on Build GRCh38
rs373557692 Chr.20: 64198968-64198968 on Build GRCh38
rs746171306 Chr.20: 64198855-64198855 on Build GRCh38
rs540037019 Chr.20: 64198892-64198892 on Build GRCh38

View Full Product Details